Merck
  • Diagnosis of common bacterial causes of urethritis in men by Gram stain, culture and multiplex PCR.
Diagnosis of common bacterial causes of urethritis in men by Gram stain, culture and multiplex PCR.
The Malaysian journal of pathology (2014-12-17)
F Jahan, S M Shamsuzzaman, S Akter
ABSTRACT
Urethritis is one of the most important causes of morbidity and mortality in developing countries. The aim of this study was to detect common bacterial causes of urethritis in men by Gram stain, culture and multiplex PCR.185 male patients who presented at the Skin and venereal clinic of the Dhaka Medical College, Bangladesh with clinical symptoms suggestive of urethritis were enrolled in this study. Urethral discharges were tested for detection of Neisseria gonorrhoeae by Gram stain, culture and PCR. Multiplex PCR assay was done to detect DNA of Chlamydia trachomatis, Ureaplasma urealyticum and Mycoplasma genitalium. Out of 185 participants, 30.27% and 14.6% were infected by Neisseria gonorrhoeae and Chlamydia trachomatis respectively. None of the individuals was found positive for either Ureaplasma urealyticum or Mycoplasma genitalium. Among the Neisseria gonorrhoeae positive patients 27.57% were positive from Gram stain, 26.49% were culture positive, 30.27% were positive by PCR (p<0.001). 32.65% of the Neisseria gonorrhoeae isolates were penicillinase producers and 83.67% were susceptible to ceftriaxone. Considering culture as the gold standard, the sensitivity and specificity of PCR for the detection of Neisseria gonorrhoeae was 100%, and 94.85% respectively with an accuracy of 96.22%. 3.73% of the 134 smear negative and 5.15% of the 136 culture negative samples were positive by PCR. PCR was the most sensitive and rapid method for the diagnosis of urethritis. Multiplex PCR may be a useful approach to laboratory diagnosis of urethritis in men for its high sensitivity and specificity.
Sigma-Aldrich
Crystal Violet, certified by the BSC
Sigma-Aldrich
Crystal Violet Solution
Sigma-Aldrich
Crystal Violet Solution
Supelco
Crystal Violet, VETRANAL®, analytical standard
Sigma-Aldrich
Crystal Violet, for microscopy (Bact., Bot., Hist., Vit.), indicator (pH 0.1-2.0)
Sigma-Aldrich
Crystal Violet, ACS reagent, ≥90.0% anhydrous basis
Sigma-Aldrich
Crystal Violet, S. No.: 785
Methylrosanilinium for system suitability, European Pharmacopoeia (EP) Reference Standard
Methylrosanilinium chloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Crystal Violet Solution, 1%, aqueous solution
Sigma-Aldrich
Gentian Violet, meets USP testing specifications