Merck
  • Carfilzomib, lenalidomide, and dexamethasone for relapsed multiple myeloma.
Carfilzomib, lenalidomide, and dexamethasone for relapsed multiple myeloma.
The New England journal of medicine (2014-12-09)
A Keith Stewart, S Vincent Rajkumar, Meletios A Dimopoulos, Tamás Masszi, Ivan Špička, Albert Oriol, Roman Hájek, Laura Rosiñol, David S Siegel, Georgi G Mihaylov, Vesselina Goranova-Marinova, Péter Rajnics, Aleksandr Suvorov, Ruben Niesvizky, Andrzej J Jakubowiak, Jesus F San-Miguel, Heinz Ludwig, Michael Wang, Vladimír Maisnar, Jiri Minarik, William I Bensinger, Maria-Victoria Mateos, Dina Ben-Yehuda, Vishal Kukreti, Naseem Zojwalla, Margaret E Tonda, Xinqun Yang, Biao Xing, Philippe Moreau, Antonio Palumbo
ABSTRACT
Lenalidomide plus dexamethasone is a reference treatment for relapsed multiple myeloma. The combination of the proteasome inhibitor carfilzomib with lenalidomide and dexamethasone has shown efficacy in a phase 1 and 2 study in relapsed multiple myeloma. We randomly assigned 792 patients with relapsed multiple myeloma to carfilzomib with lenalidomide and dexamethasone (carfilzomib group) or lenalidomide and dexamethasone alone (control group). The primary end point was progression-free survival. Progression-free survival was significantly improved with carfilzomib (median, 26.3 months, vs. 17.6 months in the control group; hazard ratio for progression or death, 0.69; 95% confidence interval [CI], 0.57 to 0.83; P=0.0001). The median overall survival was not reached in either group at the interim analysis. The Kaplan-Meier 24-month overall survival rates were 73.3% and 65.0% in the carfilzomib and control groups, respectively (hazard ratio for death, 0.79; 95% CI, 0.63 to 0.99; P=0.04). The rates of overall response (partial response or better) were 87.1% and 66.7% in the carfilzomib and control groups, respectively (P<0.001; 31.8% and 9.3% of patients in the respective groups had a complete response or better; 14.1% and 4.3% had a stringent complete response). Adverse events of grade 3 or higher were reported in 83.7% and 80.7% of patients in the carfilzomib and control groups, respectively; 15.3% and 17.7% of patients discontinued treatment owing to adverse events. Patients in the carfilzomib group reported superior health-related quality of life. In patients with relapsed multiple myeloma, the addition of carfilzomib to lenalidomide and dexamethasone resulted in significantly improved progression-free survival at the interim analysis and had a favorable risk-benefit profile. (Funded by Onyx Pharmaceuticals; ClinicalTrials.gov number, NCT01080391.).
Product Description
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
(±)-Thalidomide, ≥98%, powder
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethasone-Water Soluble, suitable for cell culture, BioReagent
Dexamethasone, British Pharmacopoeia (BP) Assay Standard
USP
Dexamethasone, United States Pharmacopeia (USP) Reference Standard
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard