Merck
  • Cholesteryl ester diffusion, location and self-association constraints determine CETP activity with discoidal HDL: excimer probe study.
Cholesteryl ester diffusion, location and self-association constraints determine CETP activity with discoidal HDL: excimer probe study.
Archives of biochemistry and biophysics (2014-12-03)
Alexander D Dergunov, Elena V Shabrova, Gennady E Dobretsov
ABSTRACT
The transfer of cholesteryl ester by recombinant cholesteryl ester transfer protein (CETP) between reconstituted discoidal high-density lipoprotein (rHDL) was studied. Particles contained apolipoprotein A-I, unsaturated POPC or saturated DPPC and cholesteryl ester as cholesteryl 1-pyrenedecanoate (CPD) or cholesteryl laurate (CL) in donor and acceptor rHDL, respectively. Probe dynamics fulfilled the quenching sphere-of-action model. The cholesteryl ester exchange between donor and acceptor particles was characterized by a heterogeneous kinetics; the fast exchanging CPD pool was much higher in a case of POPC compared to DPPC complexes. Probe fraction accessible to CETP increased with temperature, suggesting a more homogeneous probe distribution. Noncompetitive inhibition of probe transfer by acceptor particles was observed. The values of Vmax (0.063μMmin(-1)) and catalytic rate constant kcat (0.42s(-1)) together with a similarity of Km (0.9μM CPD) and KI (2.8μM CL) values for POPC-containing rHDL suggest the efficient cholesteryl ester transfer between nascent HDL with unsaturated phosphatidylcholine in vivo. The phospholipid matrix in discoidal HDL may underlie CETP activity through the self-association, diffusivity and location of cholesteryl ester in the bilayer, the accessibility of cholesteryl ester to cholesterol-binding site in apoA-I structure and the binding of cholesteryl ester, positionable by apoA-I, to CETP.
Product Description
Supelco
Cholesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, aqueous solution, sterile-filtered, suitable for cell culture
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥99.0% (TLC)
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
Cholesterol, from lanolin, ≥99.0% (GC)
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥95.5% (GC), ≥98% (TLC)
SAFC
Cholesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
SAFC
Cholesterol, Plant-Derived, SyntheChol®
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, ≥99% (TLC)
Sigma-Aldrich
1,2-Dipalmitoyl-sn-glycero-3-phosphocholine, semisynthetic, ≥99%