Merck
The current place of aprotinin in the management of bleeding.
Anaesthesia (2014-12-03)
D Royston
ABSTRACT
There is a considerable difference between the mechanism of action of the lysine analogues, tranexamic acid and epsilon-aminocaproic acid, and the serine protease inhibitor aprotinin. Aprotinin acts to inactivate free plasmin, but with little effect on bound plasmin, whereas the lysine analogues are designed to prevent excessive plasmin formation by fitting into plasminogen's lysine-binding site to prevent the binding of plasminogen to fibrin. Aprotinin is associated with a reduction in bleeding and transfusion requirements following major surgery, and has a dose-response profile, compared with no dose-response effect in the one study investigating tranexamic acid in cardiac surgical patients. Following its withdrawal in 2007, which is explained in detail in this review, the regulators have now licensed aprotinin for myocardial revascularisation only, which is relatively low-risk for bleeding.
Sigma-Aldrich
Aprotinin from bovine lung, BioUltra, 3-8 TIU/mg solid, ≥98% (SDS-PAGE)
Sigma-Aldrich
Aprotinin from bovine lung
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-7 TIU/mg solid
Sigma-Aldrich
Aprotinin from bovine lung, saline solution, 3-7 TIU/mg protein
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid, BioReagent, suitable for cell culture
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized powder, 3-8 TIU/mg solid
Aprotinin for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Aprotinin from bovine lung, lyophilized, ~80% (HPCE), crystalline (fine), white, ≥3500 U/mg
Sigma-Aldrich
trans-4-(Aminomethyl)cyclohexanecarboxylic acid, 97%
Aprotinin solution, BRP, European Pharmacopoeia (EP) Reference Standard
USP
Tranexamic acid, United States Pharmacopeia (USP) Reference Standard
Tranexamic acid, European Pharmacopoeia (EP) Reference Standard