Merck
  • Ibritumomab consolidation after 3 cycles of CHOP plus radiotherapy in high-risk limited-stage aggressive B-cell lymphoma: SWOG S0313.
Ibritumomab consolidation after 3 cycles of CHOP plus radiotherapy in high-risk limited-stage aggressive B-cell lymphoma: SWOG S0313.
Blood (2014-11-15)
Daniel O Persky, Thomas P Miller, Joseph M Unger, Catherine M Spier, Soham Puvvada, B Dino Stea, Oliver W Press, Louis S Constine, Kevin P Barton, Jonathan W Friedberg, Michael LeBlanc, Richard I Fisher
ABSTRACT
In the S0313 trial, we evaluated the impact of adding ibritumomab tiuxetan consolidation to 3 cycles of standard cyclophosphamide, doxorubicin, vincristine, and prednisone (CHOP) chemotherapy plus involved field radiotherapy (IFRT) in patients with limited-stage aggressive B-cell non-Hodgkin lymphoma (LD-NHL). Patients with at least 1 stage-modified adverse risk factor (nonbulky stage II, age >60 years, elevated lactate dehydrogenase, or World Health Organization performance status of 2) were treated with CHOP on days 1, 22, and 43, followed 3 weeks later by 40 to 50 Gy of IFRT. An ibritumomab tiuxetan regimen was initiated 3 to 6 weeks following IFRT. Forty-six patients were registered and eligible, with median follow-up of 7.3 years. The progression-free survival estimate is 89% at 2 years, 82% at 5 years, and 75% at 7 years. The overall survival estimate is 91% at 2 years, 87% at 5 years, and 82% at 7 years. Grade 4 adverse events occurring more than once included neutropenia (8), leukopenia (5), and lymphopenia (2). Febrile neutropenia was observed in 4 patients. No cases of treatment-related myeloid neoplasms were noted. In conclusion, patients with high-risk LD-NHL treated with 3 cycles of CHOP plus IFRT followed by ibritumomab tiuxetan consolidation had outcomes that compare favorably to our historical experience. The clinical trial was registered at www.clinicaltrials.gov as #NCT00070018.
Product Description
Supelco
Prednisone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Sigma-Aldrich
Prednisone, ≥98%
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard
USP
Prednisone, United States Pharmacopeia (USP) Reference Standard
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals