Merck
  • Melatonin receptors trigger cAMP production and inhibit chloride movements in nonpigmented ciliary epithelial cells.
Melatonin receptors trigger cAMP production and inhibit chloride movements in nonpigmented ciliary epithelial cells.
The Journal of pharmacology and experimental therapeutics (2014-10-26)
Fernando Huete-Toral, Almudena Crooke, Alejandro Martínez-Águila, Jesús Pintor
ABSTRACT
Melatonin and its analog 5-MCA-NAT (5-methylcarboxyamino-N-acetyl tryptamine) are active compounds reducing intraocular pressure (IOP). This action is mediated through MT2 and the putative MT3 melatonin receptor, producing a transient reduction of IOP that lasts for a few hours and has not yet been characterized. The use of melatonin and its analog are causing a decrease in chloride efflux from rabbit nonpigmented epithelial cells (NPE), possibly explaining the decrease in IOP. Melatonin and 5-MCA-NAT inhibited rabbit NPE chloride release in a concentration-dependent manner, whereas the pD2 values were between 4.5 ± 1.2 and 4.4 ± 1.0, respectively. Melatonin hypotensive action was enhanced by the presence of MT2 antagonists, such as DH97 (N-pentanoyl-2-benzyltryptamine) and 4-P-P-DOT (4-phenyl-2-propionamidotetralin) and by the nonselective melatonin receptor antagonist luzindole. Prazosin (1.5 µM) partially reverses the melatonin action by acting as a selective MT3 antagonist. However, at 15 nM it acts as an α-adrenergic receptor antagonist, enhancing the melatonin effect. Regarding the intracellular pathways triggered by melatonin receptors, neither phospholipase C/protein kinase C pathway nor the canonical reduction of intracellular cAMP was responsible for melatonin or 5-MCA-NAT actions. On the contrary, the application of these substances produced a concentration-dependent increase of cAMP, with pD2 values of 4.6 ± 0.2 and 4.9 ± 0.7 for melatonin and 5-MCA-NAT, respectively. In summary, melatonin reduces the release of chloride concomitantly to cAMP generation. The reduction of Cl(-) secretion accounts for a decrease in the water outflow and therefore a decrease in aqueous humor production. This could be one of the main mechanisms responsible for the reduction of IOP after application of melatonin and 5-MCA-NAT.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Melatonin, powder, ≥98% (TLC)
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate tris salt, ≥97% (HPLC), powder
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate, ≥98.5% (HPLC), powder
USP
Melatonin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Okadaic acid from Prorocentrum concavum, 92-100% (HPLC)
Sigma-Aldrich
Adenosine 3′,5′-cyclic monophosphate sodium salt monohydrate, ≥98.0% (HPLC), powder
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane