Merck
  • Homocysteine promotes intestinal fibrosis in rats with trinitrobenzene sulfonic acid-induced colitis.
Homocysteine promotes intestinal fibrosis in rats with trinitrobenzene sulfonic acid-induced colitis.
Digestive diseases and sciences (2014-10-09)
Hao Ding, Hui-Zhong Gan, Wen-Jie Fan, Li-Yu Cao, Jian-Ming Xu, Qiao Mei
ABSTRACT
Previous studies have revealed significantly increased levels of plasma and mucosal homocysteine (Hcy) in patients with Crohn's disease (CD); however, whether Hcy is involved in intestinal fibrosis of CD remains unclear. This study aimed to investigate the effects of Hcy on intestinal fibrosis in TNBS/ethanol-induced colitis and to elucidate its potential mechanisms. Sprague-Dawley rats were divided into 4 groups: normal control, normal + Hcy injection, TNBS model and TNBS model + Hcy injection. Hyperhomocysteinemia was induced by subcutaneous injection of Hcy. DAI, CMDI and HI were calculated to evaluate the severity of colitis. Masson trichrome staining was performed to assess the severity of fibrosis. The plasma and mucosal levels of Hcy were measured by HPLC-FD. The levels of IL-1β, IL-6, TNF-α, TGF-β1, CTGF, MMP-2,9 and collagen I, III in the colon were determined by ELISA, and the mRNA expressions of TGF-β1, MMP-2,9 and TIMP-1 were detected by RT-PCR. Hcy was found to increase the scores of DAI, CMDI and HI; levels of IL-1β, Il-6, TNF-α, TGF-β1, CTGF, MMP-2,9 and collagen I, III; and mRNA expressions of TGF-β1, MMP-2,9 and TIMP-1 in colonic tissue of rats with TNBS/ethanol-induced colitis. Hcy promotes intestinal fibrosis in rats with TNBS/ethanol-induced colitis, the underlying mechanisms of which may be attributed to its effects of increasing inflammatory damage, promoting the expression of profibrogenic cytokines and influencing MMPs/TIMPs balance.
Product Description
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Homocysteine, ≥98.0% (NT)
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.005% water
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure 190 proof, for molecular biology
Sigma-Aldrich
Picrylsulfonic acid solution, 5 % (w/v) in H2O, BioReagent, suitable for determination of primary amines
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)
Supelco
Ethanol-10, 10 mg/dL in H2O, pack of 10 × 1.2 mL ampules, certified reference material, Cerilliant®
Supelco
Ethanol-150, 150 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethyl alcohol, Pure 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Supelco
Ethanol-100 (10 ampules/kit), 100 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-200 (10 ampules/kit), 200 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-300, 300 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-50, 50 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-80, 80 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-40, 40 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)