Merck
  • Incorporation of raloxifene-impregnated allograft around orthopedic titanium implants impairs early fixation but improves new bone formation.
Incorporation of raloxifene-impregnated allograft around orthopedic titanium implants impairs early fixation but improves new bone formation.
Acta orthopaedica (2014-09-02)
Lars L Hermansen, Mette Sørensen, Jeppe Barckman, Joan E Bechtold, Kjeld Søballe, Jørgen Baas
ABSTRACT
The anti-osteoporotic drug raloxifene reduces the risk of vertebral fractures by increasing bone mass density. We investigated whether raloxifene offers any benefits in augmenting early fixation of orthopedic implants in the setting of impaction bone grafting. 24 non-weight-bearing grafted gap implants were inserted bilaterally into the tibia of 12 dogs. The 2.5-mm peri-implant gap was filled with either raloxifene-impregnated or untreated bone allograft. Implants were harvested after 28 days. Implant fixation was assessed by mechanical testing and histomorphometric evaluation. Raloxifene-treated allograft reduced early implant fixation compared to untreated allograft, as measured by inferior maximum shear strength (p < 0.001) and apparent shear stiffness (p = 0.001). We found that the raloxifene group had more newly formed bone in the gap around the implant (p = 0.02), but also less allograft (p = 0.03). The accelerated allograft resorption in the raloxifene group explained the impaired early fixation, despite its stimulation of new bone formation. Our results with local and possible high-dose treatment are not consistent with current theory regarding the mechanism of how systemic raloxifene administration counteracts the decrease in BMD in postmenopausal women. Instead of being solely anti-resorptive as generally held, our results indicate a possible anabolic side of raloxifene.
Product Description
Titanium, rod, 100mm, diameter 20mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 0.81mm, inside diameter 0.61mm, wall thickness 0.1mm, as drawn, 99.6+%
Titanium, rod, 100mm, diameter 3.0mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, mesh, 100x100mm, nominal aperture 4.3mm, wire diameter 1.5mm, 4.4x4.4 wires/inch, open area 94%, platinized diamond mesh
Titanium, sphere, 100pcs, diameter 3mm, 99.6+%
Titanium, tube, 200mm, outside diameter 6.1mm, inside diameter 5.1mm, wall thickness 0.5mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 3.0mm, inside diameter 1.5mm, wall thickness 0.75mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, rod, 50mm, diameter 8.0mm, as drawn, 99.99+%
Titanium, tube, 200mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Titanium, rod, 25mm, diameter 9.5mm, annealed, 99.99+%
Titanium, tube, 200mm, outside diameter 9.5mm, inside diameter 8.2mm, wall thickness 0.65mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 2.03mm, inside diameter 1.63mm, wall thickness 0.2mm, annealed, 99.6+%
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium, sponge, 1-20 mm, 99.5% trace metals basis
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.7% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Toluidine Blue, for microscopy (Hist., Vit.), 8.74% (ZN (THEORY))
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Titanium, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Titanium, wire, diam. 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, sputtering target, diam. × thickness 2.00 in. × 0.25 in., 99.995% trace metals basis
Raloxifene hydrochloride for peak identification, European Pharmacopoeia (EP) Reference Standard
Titanium, wire reel, 5m, diameter 0.25mm, as drawn, 99.6+%
Titanium, wire reel, 500m, diameter 0.25mm, annealed, 99.6+%