Merck
  • A new immunostain algorithm classifies diffuse large B-cell lymphoma into molecular subtypes with high accuracy.
A new immunostain algorithm classifies diffuse large B-cell lymphoma into molecular subtypes with high accuracy.
Clinical cancer research : an official journal of the American Association for Cancer Research (2009-08-27)
William W L Choi, Dennis D Weisenburger, Timothy C Greiner, Miguel A Piris, Alison H Banham, Jan Delabie, Rita M Braziel, Huimin Geng, Javeed Iqbal, Georg Lenz, Julie M Vose, Christine P Hans, Kai Fu, Lynette M Smith, Min Li, Zhongfeng Liu, Randy D Gascoyne, Andreas Rosenwald, German Ott, Lisa M Rimsza, Elias Campo, Elaine S Jaffe, David L Jaye, Louis M Staudt, Wing C Chan
ABSTRACT
Hans and coworkers previously developed an immunohistochemical algorithm with approximately 80% concordance with the gene expression profiling (GEP) classification of diffuse large B-cell lymphoma (DLBCL) into the germinal center B-cell-like (GCB) and activated B-cell-like (ABC) subtypes. Since then, new antibodies specific to germinal center B-cells have been developed, which might improve the performance of an immunostain algorithm. We studied 84 cases of cyclophosphamide-doxorubicin-vincristine-prednisone (CHOP)-treated DLBCL (47 GCB, 37 ABC) with GCET1, CD10, BCL6, MUM1, FOXP1, BCL2, MTA3, and cyclin D2 immunostains, and compared different combinations of the immunostaining results with the GEP classification. A perturbation analysis was also applied to eliminate the possible effects of interobserver or intraobserver variations. A separate set of 63 DLBCL cases treated with rituximab plus CHOP (37 GCB, 26 ABC) was used to validate the new algorithm. A new algorithm using GCET1, CD10, BCL6, MUM1, and FOXP1 was derived that closely approximated the GEP classification with 93% concordance. Perturbation analysis indicated that the algorithm was robust within the range of observer variance. The new algorithm predicted 3-year overall survival of the validation set [GCB (87%) versus ABC (44%); P < 0.001], simulating the predictive power of the GEP classification. For a group of seven primary mediastinal large B-cell lymphoma, the new algorithm is a better prognostic classifier (all "GCB") than the Hans' algorithm (two GCB, five non-GCB). Our new algorithm is significantly more accurate than the Hans' algorithm and will facilitate risk stratification of DLBCL patients and future DLBCL research using archival materials.