Merck
  • Detection of gonadotropin-releasing hormone receptor in normal human pituitary cells and pituitary adenomas using immunohistochemistry.
Detection of gonadotropin-releasing hormone receptor in normal human pituitary cells and pituitary adenomas using immunohistochemistry.
Virchows Archiv : an international journal of pathology (2000-10-19)
S La Rosa, N Celato, S Uccella, C Capella
ABSTRACT
Gonadotropin-releasing hormone (GnRH), which is a well-known regulator of gonadotroph function, has recently been considered to be a paracrine factor involved in the control of somatotroph, lactotroph, and corticotroph cells. GnRH action is initiated by binding to a specific cell surface receptor, the gonadotropin-releasing hormone receptor (GnRHR), which is expressed by follicle-stimulating hormone/luteinizing hormone (FSH/LH) cells. Using in situ hybridization techniques, GnRHR messenger ribonucleic acid (mRNA) has recently been detected in normal human anterior pituitary gland and in various pituitary adenomas, including FSH/LH-cell, growth hormone (GH)-cell, adrenocorticotropic hormone (ACTH)-cell, and null-cell adenomas. However, immunohistochemical studies indicating the specific cell distribution of GnRHR in normal pituitary cells have never been reported. The aim of the present investigation was to evaluate the immunohistochemical expression of GnRHR in different types of normal pituitary cells and related tumors. Using double-label immunohistochemical techniques on formalin-fixed and paraffin-embedded tissues and specific antibodies directed against pituitary hormones and GnRHR, we found GnRHR immunoreactivity not only in FSH/LH cells, but also in GH- and thyroid-stimulating hormone (TSH) cells. GnRHR was detected in FSH/LH-cell, GH-cell, mixed GH- and prolactin (PRL)-cell, and alpha-subunit (alpha-SU)/null-cell adenomas. The findings of this study suggest that the interaction between GnRH and GnRHR may play a role in paracrine/autocrine regulation of different types of normal pituitary cells and pituitary adenomas.
Sigma-Aldrich
TSH Rabbit Polyclonal Antibody
Sigma-Aldrich
FSH Rabbit Polyclonal Antibody
Sigma-Aldrich
TSH (EP254) Rabbit Monoclonal Primary Antibody