Merck
  • Enhancement of endotoxic shock by N-acetylmuramyl-L-alanyl-(L-seryl)-D-isoglutamine (muramyl dipeptide).
Enhancement of endotoxic shock by N-acetylmuramyl-L-alanyl-(L-seryl)-D-isoglutamine (muramyl dipeptide).
Cancer research (1979-11-01)
E E Ribi, J L Cantrell, K B Von Eschen, S M Schwartzman
PMID115579
ABSTRACT
We described elsewhere that the synergistic antitumor activity of endotoxic extracts from Re mutants of gram-negative bacteria and trehalose mycolate against guinea pig syngeneic line 10 tumor was abrogated after peptide substances accompanying these extracts had been removed. This activity could be restored by combining peptide-free endotoxin either with cell wall skeleton from Bacillus Calmette-Guérin, a polymeric mycolic acid-arabinogalactan-mucopeptide complex, or with a combination of two separate components, trehalose dimycolate and N-acetylmuramyl-L-alanyl-(L-seryl)-D-isoglutamine (MDP). We report here that when a combination of endotoxin (150 microgram) and a mixture of MDP (150 microgram) and trehalose dimycolate (150 microgram) was inoculated into established dermal tumors, a significant number of the animals died, presumably of endotoxic shock. All surviving animals suffered severe but temporary lethargy. When administered alone intradermally in the dose levels tested, none of the components caused severe lethargy or lethality. The lethal effects of 159 microgram of MDP also occurred in combination with relatively weak endotoxic products, such as Pseudomonas vaccine (Pseudogen), and these effects did not depend upon the presence of malignant tissue. Guinea pigs inoculated i.v. were even more susceptible inasmuch as the addition of as little as 6 microgram of MDP to 150 microgram of Pseudogen, itself not lethal, caused the death of 80% of the animals.
Sigma-Aldrich
Lipid A, diphosphoryl from Escherichia coli F583 (Rd mutant)