Merck
  • Thionation with the reagent combination of phosphorus pentasulfide and hexamethyldisiloxane.
Thionation with the reagent combination of phosphorus pentasulfide and hexamethyldisiloxane.
The Journal of organic chemistry (2002-08-31)
Thomas J Curphey
ABSTRACT
The combination of P4S10 and hexamethyldisiloxane efficiently converts esters, lactones, amides, lactams, and ketones to their corresponding thiono derivatives. In the presence of elemental sulfur, 3-oxoesters are converted to dithiolethiones by this reagent. Yields are comparable to or superior to those obtained with Lawesson's reagent. The method has the advantage that reagent-derived byproducts may be removed by a simple hydrolytic workup or by filtration through silica gel, rather than by chromatography, as required for Lawesson's reagent.
Sigma-Aldrich
Hexamethyldisiloxane, ≥98%