Merck
  • Xanthene derivatives increase glucose utilization through activation of LKB1-dependent AMP-activated protein kinase.
Xanthene derivatives increase glucose utilization through activation of LKB1-dependent AMP-activated protein kinase.
PloS one (2014-09-25)
Yonghoon Kwon, Parkyong Song, Jong Hyuk Yoon, Jaewang Ghim, Dayea Kim, Byungjun Kang, Taehoon G Lee, Jin-Ah Kim, Joong-Kwon Choi, In Kwon Youn, Hyeon-Kyu Lee, Sung Ho Ryu
ABSTRACT
5' AMP-activated protein kinase (AMPK) is a highly conserved serine-threonine kinase that regulates energy expenditure by activating catabolic metabolism and suppressing anabolic pathways to increase cellular energy levels. Therefore AMPK activators are considered to be drug targets for treatment of metabolic diseases such as diabetes mellitus. To identify novel AMPK activators, we screened xanthene derivatives. We determined that the AMPK activators 9H-xanthene-9-carboxylic acid {2,2,2-trichloro-1-[3-(3-nitro-phenyl)-thioureido]-ethyl}-amide (Xn) and 9H-xanthene-9-carboxylic acid {2,2,2-trichloro-1-[3-(3-cyano-phenyl)-thioureido]-ethyl}-amide (Xc) elevated glucose uptake in L6 myotubes by stimulating translocation of glucose transporter type 4 (GLUT4). Treatment with the chemical AMPK inhibitor compound C and infection with dominant-negative AMPKa2-virus inhibited AMPK phosphorylation and glucose uptake in myotubes induced by either Xn or Xc. Of the two major upstream kinases of AMPK, we found that Xn and Xc showed LKB1 dependency by knockdown of STK11, an ortholog of human LKB1. Single intravenous administration of Xn and Xc to high-fat diet-induced diabetic mice stimulated AMPK phosphorylation of skeletal muscle and improved glucose tolerance. Taken together, these results suggest that Xn and Xc regulate glucose homeostasis through LKB1-dependent AMPK activation and that the compounds are potential candidate drugs for the treatment of type 2 diabetes mellitus.
Sigma-Aldrich
Dorsomorphin, ≥98% (HPLC)
Sigma-Aldrich
MISSION® esiRNA, targeting human TM4SF1
Sigma-Aldrich
MISSION® esiRNA, targeting human RPL6
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Tm4sf1
Sigma-Aldrich
Anti-Myc Tag Antibody, clone 4A6, clone 4A6, Upstate®, from mouse
Sigma-Aldrich
AMPK Inhibitor, Compound C, AMPK Inhibitor, Compound C, CAS 866405-64-3, is a cell-permeable compound that inhibits KDR/VEGFR2, ALK2/BMPR-I, and AMPK kinase activities (IC₅₀ = 25.1, 148, and 234.6 nM, respectively).
Sigma-Aldrich
Anti-phospho-Acetyl CoA Carboxylase (Ser79) Antibody, Upstate®, from rabbit
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
1,1-Dimethylbiguanide hydrochloride, 97%
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Rpl6