Merck
  • Calcium-independent phospholipase A₂, group VIA, is critical for RPE cell survival.
Calcium-independent phospholipase A₂, group VIA, is critical for RPE cell survival.
Molecular vision (2014-05-03)
Miriam Kolko, Rupali Vohra, Barbro Westlund van der Burght, Kristian Poulsen, Mogens Holst Nissen
ABSTRACT
To investigate the significance of calcium-independent phospholipase A₂, group VIA (iPLA2-VIA), in RPE cell survival following responses to sodium iodate (SI) in cell cultures. The human retinal pigment epithelium (RPE) cell line (ARPE-19) cells and primary mouse-RPE cultures were treated with SI to induce cell death. Cells were transfected with an iPLA₂-VIA promoter-luciferase construct to evaluate the regulation of iPLA-VIA after exposure to SI. PCR analysis, western blot analysis, and activity assays were performed to evaluate the mRNA level, protein level, and activity levels of iPLA₂-VIA after SI exposure. Inhibitors of iPLA₂-VIA were used to explore a potential protective role in cells exposed to SI. Primary RPE cell cultures were grown from iPLA₂-VIA knockout mice and wild-type mice. The cultures were exposed to SI to investigate a possible increased protection against SI in iPLA₂-VIA knockout mice compared to wild-type mice. The study revealed upregulation of iPLA₂-VIA expression (promoter activity, iPLA₂-VIA mRNA, iPLA₂-VIA protein, and iPLA₂-VIA protein activity) in ARPE-19 cells exposed to SI. SI-induced cell death was shown to be inhibited by iPLA₂-VIA-specific inhibitors in ARPE-19 cell cultures. RPE cultures from iPLA₂-VIA knockout mice were less vulnerable to SI-induced cell death compared to RPE cultures from wild-type mice. SI -induced RPE cell death involves iPLA₂-VIA upregulation and activation, and amelioration of SI-induced RPE cell death can be facilitated by inhibitors of iPLA₂-VIA. Thus, we suggest iPLA₂-VIA as a possible pharmaceutical target to treat RPE-related retinal diseases.
Sigma-Aldrich
Arachidonyl trifluoromethyl ketone, oil, ≥97% (NMR)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, ≥99.5% (T)
SAFC
HEPES
SAFC
HEPES
Supelco
HEPES, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
2-Heptanone, ≥98%, FCC, FG
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Trypsin from porcine pancreas, lyophilized powder, BioReagent, suitable for cell culture, 1,000-2,000 BAEE units/mg solid
Sigma-Aldrich
Triton X-100, laboratory grade
Supelco
2-Heptanone, analytical standard
Sigma-Aldrich
2-Heptanone, 99%
Sigma-Aldrich
2-Heptanone, natural, 98%, FG