Merck
  • SOX9 accelerates ESC differentiation to three germ layer lineages by repressing SOX2 expression through P21 (WAF1/CIP1).
SOX9 accelerates ESC differentiation to three germ layer lineages by repressing SOX2 expression through P21 (WAF1/CIP1).
Development (Cambridge, England) (2014-11-06)
Kohei Yamamizu, David Schlessinger, Minoru S H Ko
ABSTRACT
Upon removal of culture conditions that maintain an undifferentiated state, mouse embryonic stem cells (ESCs) differentiate into various cell types. Differentiation can be facilitated by forced expression of certain transcription factors (TFs), each of which can generally specify a particular developmental lineage. We previously established 137 mouse ESC lines, each of which carried a doxycycline-controllable TF. Among them, Sox9 has unique capacity: its forced expression accelerates differentiation of mouse ESCs into cells of all three germ layers. With the additional use of specific culture conditions, overexpression of Sox9 facilitated the generation of endothelial cells, hepatocytes and neurons from ESCs. Furthermore, Sox9 action increases formation of p21 (WAF1/CIP1), which then binds to the SRR2 enhancer of pluripotency marker Sox2 and inhibits its expression. Knockdown of p21 abolishes inhibition of Sox2 and Sox9-accelerated differentiation, and reduction of Sox2 2 days after the beginning of ESC differentiation can comparably accelerate mouse ESC formation of cells of three germ layers. These data implicate the involvement of the p21-Sox2 pathway in the mechanism of accelerated ESC differentiation by Sox9 overexpression. The molecular cascade could be among the first steps to program ESC differentiation.
Sigma-Aldrich
Monoclonal ANTI-FLAG® M2 antibody produced in mouse, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Monoclonal Anti-β-Actin antibody produced in mouse, clone AC-15, ascites fluid
Sigma-Aldrich
Anti-GABA antibody produced in rabbit, affinity isolated antibody, buffered aqueous solution
Sigma-Aldrich
Monoclonal Anti-MAP2 antibody produced in mouse, clone HM-2, ascites fluid
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)
Sigma-Aldrich
Propidium iodide, ≥94% (HPLC)
Sigma-Aldrich
Anti-Cytokeratin 7 Antibody, clone RCK105, clone RCK105, Chemicon®, from mouse
Sigma-Aldrich
Anti-HNF3β/FOXA2 Antibody, Upstate®, from rabbit
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Sodium DL-lactate solution, synthetic, syrup, BioXtra, suitable for mouse embryo cell culture, 60 % (w/w)
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sodium DL-lactate solution, syrup, 60 % (w/w), synthetic
Sigma-Aldrich
Sodium DL-lactate solution, BioReagent, syrup, 60 % (w/w), synthetic, suitable for cell culture
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Sodium DL-lactate, ReagentPlus®, ≥99.0% (NT)
Sigma-Aldrich
Phenol Red, ACS reagent
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethanol, BioUltra, for molecular biology, ≥99.8%, (absolute alcohol, without additive, A15 o1)