Merck
  • Syncytin proteins incorporated in placenta exosomes are important for cell uptake and show variation in abundance in serum exosomes from patients with preeclampsia.
Syncytin proteins incorporated in placenta exosomes are important for cell uptake and show variation in abundance in serum exosomes from patients with preeclampsia.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology (2014-05-09)
Amandine Vargas, Shufeng Zhou, Maude Éthier-Chiasson, Denis Flipo, Julie Lafond, Caroline Gilbert, Benoit Barbeau
ABSTRACT
Exosomes are extracellular vesicles that mediate intercellular communication and are involved in several biological processes. The objective of our study was to determine whether endogenous retrovirus group WE, member l (ERVWE1)/syncytin-1 and endogenous retrovirus group FRD, member 1 (ERVFRDE1)/syncytin-2, encoded by human endogenous retrovirus (HERV) envelope (env) genes, are present at the surface of exosomes produced by placenta-derived villous cytotrophoblasts and whether they play a role in cellular uptake of exosomes. In addition, we sought to determine whether these proteins are present in various abundances in serum-derived exosomes from normal pregnant women vs. women with preeclampsia (PE). Isolated exosomes were analyzed for their content by Western blot, a bead-associated flow cytometry approach, and a syncytin-2 ELISA. Binding and uptake were tested through confocal and electron microscopy using the BeWo choriocarcinoma cell line. Quality control of exosome preparations consisted of detection of exosomal and nonexosomal markers. Exosome-cell interactions were compared between cells incubated in the presence of control exosomes, syncytin-1 or syncytin-2-deprived exosomes, or exosomes solely bearing the uncleaved forms of these HERV env proteins. From our data, we conclude that villous cytotrophoblast exosomes are positive for both env proteins and are rapidly taken up by BeWo cells in a syncytin-1- and syncytin-2-dependent manner and that syncytin-2 is reduced in serum-derived exosomes from women with PE when compared to exosomes from normal pregnant women.
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Acetylthiocholine chloride, ≥99% (TLC), powder
Sigma-Aldrich
L-Glutamine solution, 200 mM, solution, sterile-filtered, BioXtra, suitable for cell culture
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
PKH67 Green Fluorescent Cell Linker Midi Kit for General Cell Membrane Labeling, Distributed for Phanos Technologies
SAFC
L-Glutamine
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Deoxyribonuclease I from bovine pancreas, Type IV, lyophilized powder, ≥2,000 Kunitz units/mg protein
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Glutamine
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Anti-Cytokeratin 7 Antibody, clone LP5K, FITC conjugated, clone LP5K, Chemicon®, from mouse
Sigma-Aldrich
Furin Inhibitor I, Furin Inhibitor I, is a peptidyl chloromethylketone that binds to the catalytic site of furin and blocks its activity.
Supelco
Forskolin, analytical standard