Merck
  • Adhesive barrier/directional controlled release for cartilage repair by endogenous progenitor cell recruitment.
Adhesive barrier/directional controlled release for cartilage repair by endogenous progenitor cell recruitment.
Biomaterials (2014-12-04)
Jong Min Lee, Ji Hyun Ryu, Eun Ah Kim, Seongyeon Jo, Byung-Soo Kim, Haeshin Lee, Gun-Il Im
ABSTRACT
A new design concept in controlled release chemistry is reported in this study. Unlike current depots that release drugs in all direction by an isotropic way, we demonstrate that directional release only to a clinically beneficial direction results in improved disease treatment. To achieve the directional drug release, catecholamine adhesion chemistry was used to establish robust interfacial adhesion. For this purpose, water-resistant catechol-conjugated chitosan (CHI-C) adhesive gel patch was used. We chose a cartilage repair model to test our hypothesis. The adhesive barrier exhibited directional release of platelet-derived growth factor-AA (PDGF-AA) only toward the marrow cavity defect areas. This directional PDGF-AA release greatly promoted effective recruitment of human mesenchymal stem cell (hMSCs). Moreover, the adhesive barrier prevented further migration and dispersion of the hMSCs that otherwise were not properly located to the disease site. In vivo imaging and macroscopic histological assessments demonstrated significant improvement of cartilage tissue, suggesting directional controlled release can be a general concept for improvement of tissue regeneration. This CHI-C barrier is expected to make a significant contribution in cartilage tissue engineering without cell transplantation as well as application for other tissue engineering.
Product Description
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
CellVue® Claret Far Red Fluorescent Cell Linker Mini Kit for General Membrane Labeling, Distributed for Phanos Technologies
Sigma-Aldrich
CellVue® Claret Far Red Fluorescent Cell Linker Midi Kit for General Membrane Labeling, Distributed for Phanos Technologies
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Sigma-Aldrich
D-Luciferin, synthetic, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
D-Luciferin, synthetic
Sigma-Aldrich
Formaldehyde solution, for molecular biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehyde solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
Sigma-Aldrich
Hydrocinnamic acid, 99%
SAFC
Formaldehyde solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Supelco
Formaldehyde solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Formaldehyde solution, tested according to Ph. Eur.
Sigma-Aldrich
Formaldehyde solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Formaldehyde solution, for molecular biology, BioReagent, ≥36.0% in H2O (T)
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Fluorescein, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, for molecular biology
Sigma-Aldrich
Ethanol, purum, fine spirit, denaturated with 4.8% methanol, F25 METHYL1, ~96% (based on denaturant-free substance)
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, for molecular biology
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, meets USP testing specifications
Sigma-Aldrich
Ethyl alcohol, Pure, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
3-Phenylpropionic acid, 99%, FG
Sigma-Aldrich
3,4-Dihydroxyhydrocinnamic acid, 98%
Sigma-Aldrich
Ethanol, for residue analysis
Sigma-Aldrich
Ethanol, tested according to Ph. Eur.
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethyl alcohol, Pure, 190 proof, ACS spectrophotometric grade, 95.0%