Merck
  • Resveratrol augments paclitaxel treatment in MDA-MB-231 and paclitaxel-resistant MDA-MB-231 breast cancer cells.
Resveratrol augments paclitaxel treatment in MDA-MB-231 and paclitaxel-resistant MDA-MB-231 breast cancer cells.
Anticancer research (2014-10-03)
Alyssa A Sprouse, Brittney-Shea Herbert
ABSTRACT
Resveratrol (RES) inhibits cell growth, induces apoptosis and augments chemotherapeutics in multiple cancer types, although its effects on drug-resistant cancer cells are unknown. To study the effects of resveratrol in triple-negative breast cancer cells that are resistant to the common cancer drug, paclitaxel, a novel paclitaxel-resistant cell line was generated from the MDA-MB-231 cell line. The resistant MDA-MB-231/PacR cells exhibited a 12-fold increased resistance to paclitaxel. RES treatment reduced cell proliferation and colony formation and increased senescence and apoptosis in both parental and resistant cells. Importantly, RES augmented the effects of paclitaxel in both cell lines. Up-regulation of the MDR1 and CYP2C8 genes were shown to be potential mechanisms of paclitaxel resistance in the resistant cells. RES, both alone and in combination with paclitaxel, may be useful in the treatment of paclitaxel-sensitive and paclitaxel-resistant triple-negative breast cancer cells.
Product Description
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
Resveratrol, ≥99% (HPLC)
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Supelco
Resveratrol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Paclitaxel natural for peak identification, European Pharmacopoeia (EP) Reference Standard
Resveratrol, European Pharmacopoeia (EP) Reference Standard
Supelco
Resveratrol, analytical standard
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard
Paclitaxel, European Pharmacopoeia (EP) Reference Standard