Merck
  • Inhibition of the nucleotide-binding domain, leucine-rich containing family, pyrin-domain containing 3 inflammasome reduces the severity of experimentally induced acute pancreatitis in obese mice.
Inhibition of the nucleotide-binding domain, leucine-rich containing family, pyrin-domain containing 3 inflammasome reduces the severity of experimentally induced acute pancreatitis in obese mice.
Translational research : the journal of laboratory and clinical medicine (2014-08-26)
Jason M York, Karla J Castellanos, Robert J Cabay, Giamila Fantuzzi
ABSTRACT
Acute pancreatitis (AP), although most often a mild and self-limiting inflammatory disease, worsens to a characteristically necrotic severe acute pancreatitis (SAP) in about 20% of cases. Obesity, affecting more than one-third of American adults, is a risk factor for the development of SAP, but the exact mechanism of this association has not been identified. Coincidental with chronic low-grade inflammation, activation of the nucleotide-binding domain, leucine-rich containing family, pyrin-domain containing 3 (NLRP3) inflammasome increases with obesity. Lean mice genetically deficient in specific components of the NLRP3 inflammasome are protected from experimentally induced AP, indicating a direct involvement of this pathway in AP pathophysiology. We hypothesized that inhibition of the NLRP3 inflammasome with the sulfonylurea drug glyburide would reduce disease severity in obese mice with cerulein-induced SAP. Treatment with glyburide led to significantly reduced relative pancreatic mass and water content and less pancreatic damage and cell death in genetically obese ob/ob mice with SAP compared with vehicle-treated obese SAP mice. Glyburide administration in ob/ob mice with cerulein-induced SAP also resulted in significantly reduced serum levels of interleukin 6, lipase, and amylase and led to lower production of lipopolysaccharide-stimulated interleukin 1β release in cultured peritoneal cells, compared with vehicle-treated ob/ob mice with SAP. Together, these data indicate involvement of the NLRP3 inflammasome in obesity-associated SAP and expose the possible utility of its inhibition in prevention or treatment of SAP in obese individuals.
Product Description
Glibenclamide for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Glyburide (Glibenclamide), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
Sigma-Aldrich
Glyburide, meets USP testing specifications
Sigma-Aldrich
Glybenclamide, ≥99% (HPLC)
Glybenclamide, European Pharmacopoeia (EP) Reference Standard