Merck
  • Survival of Escherichia coli cells on solid copper surfaces is increased by glutathione.
Survival of Escherichia coli cells on solid copper surfaces is increased by glutathione.
Applied and environmental microbiology (2014-09-07)
Cornelia Große, Grit Schleuder, Christin Schmole, Dietrich H Nies
ABSTRACT
Bacteria are rapidly killed on solid copper surfaces, so this material could be useful to limit the spread of multiple-drug-resistant bacteria in hospitals. In Escherichia coli, the DNA-protecting Dps protein and the NADH:ubiquinone oxidoreductase II Ndh were not involved in tolerance to copper ions or survival on solid copper surfaces. Decreased copper tolerance under anaerobic growth conditions in the presence of ascorbate and with melibiose as the carbon source indicated that sodium-dependent symport systems may provide an import route for Cu(I) into the cytoplasm. Glutathione-free ΔcopA ΔgshA double mutants of E. coli were more rapidly inactivated on solid copper surfaces than glutathione-containing wild-type cells. Therefore, while DNA protection by Dps was not required, glutathione was needed to protect the cytoplasm and the DNA against damage mediated by solid copper surfaces, which may explain the differences in the molecular mechanisms of killing between glutathione-containing Gram-negative and glutathione-free Gram-positive bacteria.
Sigma-Aldrich
Copper, turnings, purum p.a., ≥99.0%
Sigma-Aldrich
Copper, beads, 2-8 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.5 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.25 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, powder, <75 μm, 99%
Sigma-Aldrich
Copper, foil, thickness 1.0 mm, 99.999% trace metals basis
Continuous cast copper (O), BCR®, certified reference material, rod
Sigma-Aldrich
Copper, foil, thickness 0.025 mm, 99.98% trace metals basis
Sigma-Aldrich
Copper, beads, 2-8 mm, 99.9995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 0.64 mm, 99.995% trace metals basis
Sigma-Aldrich
Copper, wire, diam. 1.0 mm, ≥99.9%
Sigma-Aldrich
Copper, wire, diam. 2.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Copper, foil, ≥99.8% (complexometric)
Sigma-Aldrich
Glutathione Assay Kit, sufficient for 700 assays
Copper, tube, 1000mm, outside diameter 6.0mm, inside diameter 4mm, wall thickness 1.0mm, as drawn, 99.9%
Copper, rod, 50mm, diameter 6.35mm, as drawn, 99.999%
Copper, tube, 1000mm, outside diameter 0.75mm, inside diameter 0.23mm, wall thickness 0.26mm, as drawn, 99.9%
Copper, sphere, 1000pcs, diameter 3.0mm, precision sphere grade 200
Copper, tube, 100mm, outside diameter 0.75mm, inside diameter 0.23mm, wall thickness 0.26mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 50mm disks, thickness 0.010mm, hard, 99.95+%
Copper - O.F.H.C., foil, 25mm disks, thickness 0.075mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 25mm disks, thickness 1.0mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 4mm disks, thickness 0.010mm, hard, 99.95+%
Copper - O.F.H.C., foil, 4mm disks, thickness 0.025mm, hard, 99.95+%
Copper - O.F.H.C., foil, 1m coil, thickness 0.010mm, hard, 99.95+%
Copper - O.F.H.C., foil, 10mm disks, thickness 0.1mm, hard, 99.95+%
Copper, tube, 500mm, outside diameter 6.0mm, inside diameter 4mm, wall thickness 1.0mm, as drawn, 99.9%
Copper - O.F.H.C., foil, 5m coil, thickness 0.075mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 1m coil, thickness 0.1mm, half hard, 99.95+%
Copper - O.F.H.C., foil, 50mm disks, thickness 0.05mm, hard, 99.95+%