Merck
  • Development and validation of a liquid chromatographic method for the stability study of a pharmaceutical formulation containing voriconazole using cellulose tris(4-chloro-3-methylphenylcarbamate) as chiral selector and polar organic mobile phases.
Development and validation of a liquid chromatographic method for the stability study of a pharmaceutical formulation containing voriconazole using cellulose tris(4-chloro-3-methylphenylcarbamate) as chiral selector and polar organic mobile phases.
Journal of chromatography. A (2014-07-19)
Anne-Catherine Servais, Radu Moldovan, Elena Farcas, Jacques Crommen, Isabelle Roland, Marianne Fillet
ABSTRACT
The ophthalmic solution of voriconazole, i.e. (2R,3S)-2-(2,4-difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1H-1,2,4-triazol-1-yl)butan-2-ol, made from an injection formulation which also contains sulfobutylether-β-cyclodextrin sodium salt as an excipient (Vfend), is used for the treatment of fungal keratitis. A liquid chromatographic (LC) method using polar organic mobile phase and cellulose tris(4-chloro-3-methylphenylcarbamate) coated on silica as chiral stationary phase was successfully developed to evaluate the chiral stability of the ophthalmic solution. The percentage of methanol (MeOH) in the mobile phase containing acetonitrile (ACN) as the main solvent significantly influenced the retention and resolution of voriconazole and its enantiomer ((2S,3R)-2-(2,4-difluorophenyl)-3-(5-fluoropyrimidin-4-yl)-1-(1H-1,2,4-triazol-1-yl)butan-2-ol). The optimized mobile phase consisted of ACN/MeOH/diethylamine/trifluoroacetic acid (80/20/0.1/0.1; v/v/v/v). The method was found to be selective not only regarding the enantiomer of voriconazole but also regarding the specified impurities described in the monograph from the European Pharmacopoeia. The LC method was then fully validated applying the strategy based on total measurement error and accuracy profiles. Under the selected conditions, the determination of 0.1% of voriconazole enantiomer could be performed. Finally, a stability study of the ophthalmic solution was conducted using the validated LC method.
Product Description
Supelco
Avicel® PH-101, ~50 μm particle size
Sigma-Aldrich
α-Cellulose, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Sigmacell Cellulose, Type 20, 20 μm
Sigma-Aldrich
Sigmacell Cellulose, Type 101, Highly purified, fibers
Sigma-Aldrich
Sigmacell Cellulose, Type 50, 50 μm
Sigma-Aldrich
α-Cellulose, powder
Supelco
Cellulose, DFS-0, microcrystalline, suitable for thin layer chromatography (TLC)
Supelco
Cellulose, DS-0, powder, suitable for thin layer chromatography (TLC)
Sigma-Aldrich
Avicel® PH-101, tested according to Ph. Eur.
Supelco
Cellulose, powder, for column chromatography
Supelco
Cellulose, acid washed, from spruce, for column chromatography
Sigma-Aldrich
Cellulose, microcrystalline, powder, 20 μm
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Cellulose, microcrystalline, powder
Sigma-Aldrich
Cellulose, colloidal, microcrystalline
Sigma-Aldrich
Cellulose, fibers, (medium)
Supelco
Cellulose, acid washed, powder, for column chromatography
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing
Sigma-Aldrich
Trifluoroacetic acid, suitable for HPLC, ≥99.0%