Merck
  • Characterization of full-length recombinant human Proteoglycan 4 as an ocular surface boundary lubricant.
Characterization of full-length recombinant human Proteoglycan 4 as an ocular surface boundary lubricant.
Experimental eye research (2014-07-06)
Michael L Samsom, Sheila Morrison, Nemanja Masala, Benjamin D Sullivan, David A Sullivan, Heather Sheardown, Tannin A Schmidt
ABSTRACT
Proteoglycan 4 (PRG4, or lubricin) is a lubricating mucin-like glycoprotein recently discovered at the ocular surface, where it functions as a boundary lubricant and appears to play a protective role. Recent technological advances have enabled abundant expression of full-length recombinant human PRG4 (rhPRG4). The objectives of this study were to 1) biochemically characterize the gross structure and glycosylations of full-length rhPRG4, and 2) assess the ocular surface boundary lubricating ability of rhPRG4 at both human cornea-eyelid and human cornea-polydimethylsiloxane (PDMS) biointerfaces. rhPRG4 expressed by a Chinese hamster ovary cell line was characterized and compared to native bovine PRG4 by SDS-PAGE western blotting, and protein identity was assessed by tandem mass spectrometry (MS/MS). Human corneas were articulated against PDMS or human eyelids, at effective sliding velocities of 0.3-30 mm/s under physiological loads of ∼15 kPa, to assess and compare the ocular lubricating ability of rhPRG4 to PRG4. Samples were tested serially in PRG4, rhPRG4 (both 300 μg/ml), then saline. Western blotting indicated that rhPRG4 had immunoreactivity at the appropriate apparent molecular weight, and possessed O-linked glycosylation consistent with that of PRG4. rhPRG4 protein identity was confirmed by MS/MS. Both PRG4 and rhPRG4 significantly, and similarly, reduced friction compared to saline at both human cornea - PDMS and human cornea-eyelid biointerfaces. In conclusion, the rhPRG4 studied here demonstrated appropriate higher order structure, O-linked glycosylations, and ocular surface boundary lubricating. Purified rhPRG4 may have clinical utility as a topical treatment of dry eye disease or contact lens biomaterial coating to promote more comfortable wear.
Product Description
Silicone oil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Silicone oil, viscosity 100,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 1,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 500 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 50 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 100 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 60,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 20 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 350 cSt (25 °C)
Sigma-Aldrich
Silicone oil, for oil baths (from −50°C to +200°C)
Sigma-Aldrich
Silicone oil, viscosity 5 cSt (25 °C)
Silicone oil, for melting point and boiling point apparatuses
Sigma-Aldrich
Silicone oil, viscosity 30,000 cSt (25 °C)
Sigma-Aldrich
Silicone oil, viscosity 10 cSt (25 °C)