Merck
  • RSK2 regulates endocytosis of FGF receptor 1 by phosphorylation on serine 789.
RSK2 regulates endocytosis of FGF receptor 1 by phosphorylation on serine 789.
Oncogene (2013-10-22)
B Nadratowska-Wesolowska, E M Haugsten, M Zakrzewska, P Jakimowicz, Y Zhen, D Pajdzik, J Wesche, A Wiedlocha
ABSTRACT
FGFR1 (fibroblast growth factor receptor 1) regulates many key cellular responses including proliferation, migration and differentiation through activation of signaling pathways. Irregularities in FGFR1 signaling have been implicated in several pathological conditions, including human cancer. In order to discover novel regulators of FGFR1 signaling, we performed yeast two-hybrid screens and identified RSK2 (p90 ribosomal S6 kinase 2) as a potential FGFR1 interaction partner. RSK2 belongs to the family of serine/threonine kinases that are activated through the Ras-MAPK signal transduction pathway. Both in vitro and in vivo experiments confirmed the interaction and we show that phosphorylated RSK2 binds to and phosphorylates serine 789 in the C-terminal tail of FGFR1. Inhibition of RSK2 activity led to prolonged tyrosine transphosphorylation of FGFR1. Furthermore, prevention of FGFR1 phosphorylation by inhibition of RSK2 activity or mutation of serine 789 to alanine reduced FGFR1 endocytosis and ubiquitination explaining mechanistically the prolonged signaling activity. We propose a novel regulatory mechanism whereby activated RSK2 directly interacts with and phosphorylates FGFR1, thereby modulating receptor signaling through regulation of endocytosis.
Sigma-Aldrich
EGF from mouse, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Serine, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
EGF human, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
hEGF, EGF, recombinant, expressed in E. coli, lyophilized powder, suitable for cell culture
Sigma-Aldrich
EGF from mouse, Animal-component free, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC)
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
L-Serine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
EGF from rat, recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture
Sigma-Aldrich
Anti-phospho-RPS6KA1 (pSer352) antibody produced in rabbit, affinity isolated antibody
Supelco
L-Serine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Fibroblast Growth Factor from bovine pituitary, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Trifluoroacetic acid, ReagentPlus®, 99%
Supelco
Trifluoroacetic acid, analytical standard
Sigma-Aldrich
Trifluoroacetic acid, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
Trifluoroacetic acid, ≥99%, for protein sequencing