Merck
  • One-pot syntheses of novel pyrazole-containing bisphosphonate esters at room temperature.
One-pot syntheses of novel pyrazole-containing bisphosphonate esters at room temperature.
Organic & biomolecular chemistry (2012-08-21)
Haoyue Xiang, Xueyu Qi, Yuyuan Xie, Guangyu Xu, Chunhao Yang
ABSTRACT
An efficient general synthetic approach has been developed to synthesize pyrazole-containing bisphosphonate (N-BPs) esters from chromenone derivatives via a sequential two-step reaction in one pot at ambient temperature in good to excellent yields. This protocol provides a new convenient method to prepare lipophilic bisphosphonate precursors with potential activities.