Merck
  • Regulation of C. elegans fat uptake and storage by acyl-CoA synthase-3 is dependent on NR5A family nuclear hormone receptor nhr-25.
Regulation of C. elegans fat uptake and storage by acyl-CoA synthase-3 is dependent on NR5A family nuclear hormone receptor nhr-25.
Cell metabolism (2010-10-05)
Brendan C Mullaney, Raymond D Blind, George A Lemieux, Carissa L Perez, Ida C Elle, Nils J Faergeman, Marc R Van Gilst, Holly A Ingraham, Kaveh Ashrafi
ABSTRACT
Acyl-CoA synthases are important for lipid synthesis and breakdown, generation of signaling molecules, and lipid modification of proteins, highlighting the challenge of understanding metabolic pathways within intact organisms. From a C. elegans mutagenesis screen, we found that loss of ACS-3, a long-chain acyl-CoA synthase, causes enhanced intestinal lipid uptake, de novo fat synthesis, and accumulation of enlarged, neutral lipid-rich intestinal depots. Here, we show that ACS-3 functions in seam cells, epidermal cells anatomically distinct from sites of fat uptake and storage, and that acs-3 mutant phenotypes require the nuclear hormone receptor NHR-25, a key regulator of C. elegans molting. Our findings suggest that ACS-3-derived long-chain fatty acyl-CoAs, perhaps incorporated into complex ligands such as phosphoinositides, modulate NHR-25 function, which in turn regulates an endocrine program of lipid uptake and synthesis. These results reveal a link between acyl-CoA synthase function and an NR5A family nuclear receptor in C. elegans.
Sigma-Aldrich
ISOGRO®-D Powder -Growth Medium, 97-99 atom % D
Sigma-Aldrich
ISOGRO®-15N,D Powder -Growth Medium, 98 atom % 15N, 97 atom % D
Sigma-Aldrich
ISOGRO®-15N Powder -Growth Medium, 98 atom % 15N
Sigma-Aldrich
ISOGRO®-13C,15N Powder -Growth Medium, 98 atom % 15N, 99 atom % 13C
Sigma-Aldrich
ISOGRO®-13C Powder -Growth Medium, 99 atom % 13C
Sigma-Aldrich
ISOGRO®-13C,15N,D Powder -Growth Medium, 98 atom % 15N, 97-99 atom % D, 99 atom % 13C