Merck
  • Glycan specificity of a testis-specific lectin chaperone calmegin and effects of hydrophobic interactions.
Glycan specificity of a testis-specific lectin chaperone calmegin and effects of hydrophobic interactions.
Biochimica et biophysica acta (2014-04-29)
Masafumi Sakono, Akira Seko, Yoichi Takeda, Jun-ichi Aikawa, Masakazu Hachisu, Akihiko Koizumi, Kohki Fujikawa, Yukishige Ito
ABSTRACT
Testis-specific chaperone calmegin is required for the generation of normal spermatozoa. Calmegin is known to be a homologue of endoplasmic reticulum (ER) residing lectin chaperone calnexin. Although functional similarity between calnexin and calmegin has been predicted, detailed information concerned with substrate recognition by calmegin, such as glycan specificity, chaperone function and binding affinity, are obscure. In this study, biochemical properties of calmegin and calnexin were compared using synthetic glycans and glycosylated or non-glycosylated proteins as substrates. Whereas their amino acid sequences are quite similar to each other, a certain difference in secondary structures was indicated by circular dichroism (CD) spectrum. While both of them inhibited protein heat-aggregation to a similar extent, calnexin exhibited a higher ability to facilitate protein folding. Similarly to calnexin, calmegin preferentially recognizes monoglucosylated glycans such as Glc1Man9GlcNAc2 (G1M9). While the surface hydrophobicity of calmegin was higher than that of calnexin, calnexin showed stronger binding to substrate. We reasoned that lectin activity, in addition to hydrophobic interaction, contributes to this strong affinity between calnexin and substrate. Although their similarity in carbohydrate binding specificities is high, there seems to be some differences in the mode of substrate recognition between calmegin and calnexin. Properties of calmegin as a lectin-chaperone were revealed in comparison with calnexin.
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Calcium carbonate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E170, precipitated, 98.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Guanidine hydrochloride, anhydrous, free-flowing, Redi-Dri, ≥99%
Supelco
Calcium carbonate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ultrapure Acetonitrile
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
SAFC
Guanidine hydrochloride
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Calcium carbonate, ReagentPlus®
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Calcium carbonate, ACS reagent, ≥99.0%, powder
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Calcium carbonate, 99.999% trace metals basis
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Sigma-Aldrich
Calcium chloride solution, BioUltra, for molecular biology, ~1 M in H2O
Supelco
Calcium ion solution for ISE, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, for molecular biology, ≥99.5% (AT)
Sigma-Aldrich
Calcium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Guanidine hydrochloride, for molecular biology, ≥99%
Sigma-Aldrich
Calcium chloride
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Supelco
Calcium carbonate, reference material for titrimetry, certified by BAM, ≥99.5%
Sigma-Aldrich
Calcium carbonate, BioUltra, precipitated, ≥99.0% (KT)
Sigma-Aldrich
Calcium carbonate, tested according to Ph. Eur.
Sigma-Aldrich
Calcium chloride, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis
Sigma-Aldrich
Calcium chloride, anhydrous, powder, 99.99% trace metals basis
Sigma-Aldrich
Calcium carbonate, powder, ≤50 μm particle size, 98%