Merck
A complex iron-calcium cofactor catalyzing phosphotransfer chemistry.
Science (New York, N.Y.) (2014-09-06)
Shee Chien Yong, Pietro Roversi, James Lillington, Fernanda Rodriguez, Martin Krehenbrink, Oliver B Zeldin, Elspeth F Garman, Susan M Lea, Ben C Berks
ABSTRACT
Alkaline phosphatases play a crucial role in phosphate acquisition by microorganisms. To expand our understanding of catalysis by this class of enzymes, we have determined the structure of the widely occurring microbial alkaline phosphatase PhoX. The enzyme contains a complex active-site cofactor comprising two antiferromagnetically coupled ferric iron ions (Fe(3+)), three calcium ions (Ca(2+)), and an oxo group bridging three of the metal ions. Notably, the main part of the cofactor resembles synthetic oxide-centered triangular metal complexes. Structures of PhoX-ligand complexes reveal how the active-site metal ions bind substrate and implicate the cofactor oxo group in the catalytic mechanism. The presence of iron in PhoX raises the possibility that iron bioavailability limits microbial phosphate acquisition.
Product Description
Iron, IRMM®, certified reference material, 0.5 mm wire
Sigma-Aldrich
Phosphatase, Alkaline bovine, recombinant, expressed in Pichia pastoris, ≥4000 units/mg protein
Sigma-Aldrich
Iron, chips, 99.98% trace metals basis
Sigma-Aldrich
Iron, puriss. p.a., carbonyl-Iron powder, low in magnesium and manganese compounds, ≥99.5% (RT)
Sigma-Aldrich
Iron, powder, −325 mesh, 97%
Sigma-Aldrich
Iron, foil, thickness 0.1 mm, ≥99.9% trace metals basis
Iron, foil, 6mm disks, thickness 0.004mm, 99.85%
Iron, foil, 50mm disks, thickness 0.25mm, hard, 99.5%
Iron, foil, 8mm disks, thickness 0.004mm, 99.99+%
Iron, foil, 8mm disks, thickness 0.5mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.5mm, as rolled, 99.99+%
Iron, foil, light tested, 25x25mm, thickness 0.0125mm, 99.99+%
Iron, foil, light tested, 50x50mm, thickness 0.025mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.125mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.5mm, hard, 99.5%
Iron, foil, not light tested, 100x100mm, thickness 0.005mm, 99.85%
Iron, foil, 50mm disks, thickness 0.38mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.020mm, 99.85%
Iron, foil, 5m coil, thickness 0.1mm, hard, 99.5%
Iron, foil, light tested, 50x50mm, thickness 0.038mm, hard, 99.5%
Iron, foil, 8mm disks, thickness 0.05mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.125mm, as rolled, 99.99+%
Iron, foil, not light tested, 100x100mm, thickness 0.007mm, 99.85%
Iron, foil, 6mm disks, thickness 0.15mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.01mm, 99.99+%
Iron, foil, 8mm disks, thickness 0.003mm, 99.85%
Iron, foil, 5m coil, thickness 0.20mm, hard, 99.5%
Iron, foil, not light tested, 100x100mm, thickness 0.008mm, 99.85%
Iron, foil, 5m coil, thickness 0.075mm, hard, 99.5%
Iron, foil, 6mm disks, thickness 0.008mm, 99.85%