Merck
  • Effects of magnesium on cytomorphology and enzyme activities in thyroid of rats.
Effects of magnesium on cytomorphology and enzyme activities in thyroid of rats.
Indian journal of experimental biology (2014-08-22)
Amar K Chandra, Haimanti Goswami, Pallav Sengupta
ABSTRACT
Till date knowledge regarding the effects of high dietary magnesium on thyroid gland is incomprehensive though certain epidemiological studies reported development of thyroid gland dysfunctions in people with chronic exposure to hard water (especially with high magnesium) despite sufficient iodine consumption. The present study is to explore the effects of chronic high dietary magnesium exposure on thyroid morphology and functional status. Male adult albino Wistar strain rats were treated with graded doses of magnesium sulphate (MgSO4; 0.5, 1.0 and 1.5 g %) for 60 days and changes in different thyroid parameters were investigated. Significantly stimulated thyroid peroxidase and Na(+)-K(+)-ATPase and altered idothyronine 5'-deiodinase type I activities, enhanced serum thyroxine (T4) (both total and free), total triiodothyronine (T3) and thyroid stimulating hormone with decreased free T3 levels and T3/T4 ratio (T3:T4) along with enlargement of thyroid with associated histopathological changes were observed in the treated groups. The results clearly confirm that chronic high dietary magnesium exposure causes potential thyroid disruption as reported in earlier epidemiological studies.
Sigma-Aldrich
Magnesium preparation, Highly Reactive Rieke® Metal, suspension, 2.5 g in THF
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, 37.5 mmol
Sigma-Aldrich
Magnesium, in a Sure/Seal bottle, turnings, anhydrous tetrahydrofuran 37.5 mmol
Sigma-Aldrich
Magnesium, purum, for Grignard reactions, ≥99.5%, turnings
Sigma-Aldrich
Magnesium, powder, ≥99%
Sigma-Aldrich
Magnesium, 20-230 mesh, reagent grade, 98%
Sigma-Aldrich
Magnesium, chips, 6-35 mesh, 99.98% trace metals basis
Sigma-Aldrich
Magnesium, ReagentPlus®, ribbon, ≥99% trace metals basis
Sigma-Aldrich
Magnesium, dendritic pieces, purified by distillation, 99.998% trace metals basis
Sigma-Aldrich
Magnesium, rod, diam. 6 mm, ≥99.9% trace metals basis
Sigma-Aldrich
Magnesium, turnings, reagent grade, 98%
Sigma-Aldrich
Magnesium, ribbon, ≥99.0% Mg basis
Sigma-Aldrich
L-Thyroxine, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Thyrotropic hormone from bovine pituitary
Sigma-Aldrich
L-Thyroxine, ≥98% (HPLC)
Sigma-Aldrich
Thyrotropic hormone from human pituitary, lyophilized powder, ≥6.2 IU/mg (immunoassay, non-sterile)
Sigma-Aldrich
Magnesium, wire, 127 μm diameter, 99.9% trace metals basis
Sigma-Aldrich
Magnesium, turnings, 5-25 mm, 99.95% trace metals basis
Sigma-Aldrich
Magnesium, grit, ≥99.0% (KT)
Magnesium, wire reel, 0.5m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.5m, diameter 1.1mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.25mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.4mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 0.1m, diameter 0.5mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 0.125mm, as drawn, 99.9+%
Magnesium, microleaf, 50x50mm, thinness 1.0μm, specific density 174μg/cm2, removable support, 99.9%
Magnesium, wire reel, 0.1m, diameter 1.0mm, as drawn, 99.9+%
Magnesium, wire reel, 1m, diameter 1.1mm, as drawn, 99.9+%