Merck
  • Thermohaline circulation crisis and impacts during the mid-Pleistocene transition.
Thermohaline circulation crisis and impacts during the mid-Pleistocene transition.
Science (New York, N.Y.) (2014-06-28)
Leopoldo D Pena, Steven L Goldstein
ABSTRACT
The mid-Pleistocene transition (MPT) marked a fundamental change in glacial-interglacial periodicity, when it increased from ~41-thousand-year to 100-thousand-year cycles and developed higher-amplitude climate variability without substantial changes in the Milankovitch forcing. Here, we document, by using Nd isotopes, a major disruption of the ocean thermohaline circulation (THC) system during the MPT between marine isotope stages (MISs) 25 and 21 at ~950 to 860 thousand years ago, which effectively marks the first 100-thousand-year cycle, including an exceptional weakening through a critical interglacial (MIS 23) at ~900 thousand years ago. Its recovery into the post-MPT 100-thousand-year world is characterized by continued weak glacial THC. The MPT ocean circulation crisis facilitated the coeval drawdown of atmospheric CO2 and high-latitude ice sheet growth, generating the conditions that stabilized 100-thousand-year cycles.
Sigma-Aldrich
Carbon-12C dioxide, 99.99 atom % 12C
Sigma-Aldrich
Carbon-12C dioxide, 99.9 atom % 12C
Sigma-Aldrich
Carbon dioxide, ≥99.8%
Sigma-Aldrich
Neodymium, powder, −40 mesh, ≥99% trace rare earth metals basis
Neodymium, rod, 50mm, diameter 12.5mm, cast, 99%