Merck
  • Cooperative binding of the outer arm-docking complex underlies the regular arrangement of outer arm dynein in the axoneme.
Cooperative binding of the outer arm-docking complex underlies the regular arrangement of outer arm dynein in the axoneme.
Proceedings of the National Academy of Sciences of the United States of America (2014-07-01)
Mikito Owa, Akane Furuta, Jiro Usukura, Fumio Arisaka, Stephen M King, George B Witman, Ritsu Kamiya, Ken-ichi Wakabayashi
ABSTRACT
Outer arm dynein (OAD) in cilia and flagella is bound to the outer doublet microtubules every 24 nm. Periodic binding of OADs at specific sites is important for efficient cilia/flagella beating; however, the molecular mechanism that specifies OAD arrangement remains elusive. Studies using the green alga Chlamydomonas reinhardtii have shown that the OAD-docking complex (ODA-DC), a heterotrimeric complex present at the OAD base, functions as the OAD docking site on the doublet. We find that the ODA-DC has an ellipsoidal shape ∼24 nm in length. In mutant axonemes that lack OAD but retain the ODA-DC, ODA-DC molecules are aligned in an end-to-end manner along the outer doublets. When flagella of a mutant lacking ODA-DCs are supplied with ODA-DCs upon gamete fusion, ODA-DC molecules first bind to the mutant axonemes in the proximal region, and the occupied region gradually extends toward the tip, followed by binding of OADs. This and other results indicate that a cooperative association of the ODA-DC underlies its function as the OAD-docking site and is the determinant of the 24-nm periodicity.
Product Description
Glutathione, European Pharmacopoeia (EP) Reference Standard
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Brilliant Blue G, 250, suitable for microscopy
Sigma-Aldrich
Brilliant Blue G, pure
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Brilliant Blue G solution, Concentrate