Merck
  • Chasing protons: how isothermal titration calorimetry, mutagenesis, and pKa calculations trace the locus of charge in ligand binding to a tRNA-binding enzyme.
Chasing protons: how isothermal titration calorimetry, mutagenesis, and pKa calculations trace the locus of charge in ligand binding to a tRNA-binding enzyme.
Journal of medicinal chemistry (2014-06-24)
Manuel Neeb, Paul Czodrowski, Andreas Heine, Luzi Jakob Barandun, Christoph Hohn, François Diederich, Gerhard Klebe
ABSTRACT
Drug molecules should remain uncharged while traveling through the body and crossing membranes and should only adopt charged state upon protein binding, particularly if charge-assisted interactions can be established in deeply buried binding pockets. Such strategy requires careful pKa design and methods to elucidate whether and where protonation-state changes occur. We investigated the protonation inventory in a series of lin-benzoguanines binding to tRNA-guanine transglycosylase, showing pronounced buffer dependency during ITC measurements. Chemical modifications of the parent scaffold along with ITC measurements, pKa calculations, and site-directed mutagenesis allow elucidating the protonation site. The parent scaffold exhibits two guanidine-type portions, both likely candidates for proton uptake. Even mutually compensating effects resulting from proton release of the protein and simultaneous uptake by the ligand can be excluded. Two adjacent aspartates induce a strong pKa shift at the ligand site, resulting in protonation-state transition. Furthermore, an array of two parallel H-bonds avoiding secondary repulsive effects contributes to the high-affinity binding of the lin-benzoguanines.
Sigma-Aldrich
Ribonucleic acid, transfer from Escherichia coli, Type XX, Strain W, lyophilized powder
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
SAFC
L-Aspartic acid
Aspartic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Ribonucleic acid, transfer from bovine liver, Type XI, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from wheat germ, Type V, 15-19 units/mg solid
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), Type X-SA, lyophilized powder