Merck
  • Hypericin suppresses osteoclast formation and wear particle-induced osteolysis via modulating ERK signalling pathway.
Hypericin suppresses osteoclast formation and wear particle-induced osteolysis via modulating ERK signalling pathway.
Biochemical pharmacology (2014-06-21)
Zhengxiao Ouyang, Zanjing Zhai, Haowei Li, Xuqiang Liu, Xinhua Qu, Xianan Li, Qiming Fan, Tingting Tang, An Qin, Kerong Dai
ABSTRACT
Osteoclast-induced bone resorption and wear-particle-induced osteolysis leads to prosthetic loosening, one of the most common causes of joint implant failure, resulting in revision surgery. Thus, inhibition of osteoclastic bone resorption, which further prevents wear particle-induced osteolysis, is a potential treatment strategy for prosthetic loosening. Here, we examined the therapeutic effect of hypericin (HP), which was photosensitive, on osteoclastogenesis and wear particle-induced osteolysis in the absence of visible light. HP inhibited RANKL-induced osteoclast differentiation in bone marrow macrophages (BMMs) and RAW264.7 cell line without any evidence of cytotoxicity. The bone-resorbing activity of mature osteoclasts was significantly inhibited by HP. As HP has been previously reported to inhibit signalling pathway such as ERK and NF-κB in other cells, which is also important in osteoclast differentiation. We thus examined the molecular mechanism and showed that HP significantly inhibited the ERK/mitogen-activated protein kinase (MAPK) signalling pathway without affecting nuclear factor kappaB (NF-κB), c-Jun N-terminal kinase (JNK) and p38 signalling in RANKL-stimulated BMMs. Further in vivo studies revealed HP attenuated osteoclast formation and subsequently prevented wear particle-induced bone erosion. Taken together, the results suggest that HP inhibits RANKL-mediated osteoclastogenesis via affecting ERK signalling in vitro and suppresses wear particle-induced osteolysis in vivo. We therefore conclude that HP may be an innovative and safe alternative treatment for osteoclast-related prosthetic loosening.
Sigma-Aldrich
Ser-Phe-Leu-Leu-Arg-Asn-Pro-Asn-Asp-Lys-Tyr-Glu-Pro-Phe, ≥97% (HPLC)
Sigma-Aldrich
DL-Glyceraldehyde 3-phosphate solution, 45-55 mg/mL in H2O
Hypericin, primary reference standard
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Perylene, sublimed grade, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, for molecular biology, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Perylene, ≥99%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, BioUltra, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Hypericin from Hypericum perforatum, ~95% (HPLC)
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%