Merck
  • Augmented post-remission therapy for a minimal residual disease-defined high-risk subgroup of children and young people with clinical standard-risk and intermediate-risk acute lymphoblastic leukaemia (UKALL 2003): a randomised controlled trial.
Augmented post-remission therapy for a minimal residual disease-defined high-risk subgroup of children and young people with clinical standard-risk and intermediate-risk acute lymphoblastic leukaemia (UKALL 2003): a randomised controlled trial.
The Lancet. Oncology (2014-06-14)
Ajay Vora, Nick Goulden, Chris Mitchell, Jeremy Hancock, Rachael Hough, Clare Rowntree, Anthony V Moorman, Rachel Wade
ABSTRACT
No randomised study has shown whether stratification of treatment by minimal residual disease (MRD) response improves outcome in children and young people with acute lymphoblastic leukaemia (ALL). We assessed whether children and young people with clinical standard and intermediate-risk ALL who have persistent MRD at the end of induction therapy benefit from augmented post-remission therapy. Between Oct 1, 2003, and June 30, 2011, we enrolled eligible patients aged 1-24 years and initially categorised them into clinical standard-risk, intermediate-risk, and high-risk groups on the basis of a combination of National Cancer Institute criteria, cytogenetics, and early morphological response to induction therapy. Clinical standard-risk and intermediate-risk patients with MRD of 0·01% or higher at day 29 of induction (MRD high risk) were randomly assigned (1:1) to standard therapy (treatment regimens A and B) or augmented post-remission therapy (regimen C). Compared with standard therapy, the augmented treatment regimen (regimen C) included an additional eight doses of pegylated asparaginase, 18 doses of vincristine, and escalated-dose intravenous methotrexate without folinic acid rescue during interim maintenance courses. Computer randomisation was used for treatment allocation and was balanced for sex, age (<10 years vs ≥10 years), and white blood cell count at diagnosis (<50 × 10(9)/L vs ≥50 × 10(9)/L) by minimisation. Patients, clinicians, and data analysts were not masked to treatment allocation. The primary outcomes were event-free survival and overall survival. Analyses were by intention to treat. This trial is registered with Current Controlled Trials, number ISRCTN07355119. 533 MRD high-risk patients were randomly assigned to receive standard (n=266) or augmented (n=267) post-remission therapy. After a median follow-up of 70 months (IQR 52-91), 5-year event-free survival was better in the augmented treatment group (89·6% [95% CI 85·9-93·3]) than in the standard group (82·8% [78·1-87·5]; odds ratio [OR] 0·61 [95% CI 0·39-0·98], p=0·04). Overall survival at 5 years was numerically, but not significantly, higher in the augmented treatment group (92·9% [95% CI 89·8-96·0]) than in the standard therapy group (88·9% [85·0-92·8]; OR 0·67 [95% CI 0·38-1·17], p=0·16). More adverse events occurred in the augmented treatment group than in the standard group (asparaginase-related hypersensitivity in 18 [6·7%] in the augmented group vs two [0·8%] in the standard group and asparaginase-related pancreatitis in eight [3·0%] vs one [0·4%]; intravenous methotrexate-related mucositis in 11 [4·1%] vs three [1·1%] and methotrexate-related stomatitis in 48 [18·0%] vs 12 [4·5%]). Our findings suggest that children and young people with acute lymphoblastic leukaemia and 0·01% or more MRD at the end of remission induction therapy could benefit from augmented post-remission therapy. However, the asparaginase and intravenous methotrexate used in the augmented treatment regimen is associated with more adverse events than is the standard post-remission treatment regimen. Medical Research Council and Leukaemia and Lymphoma Research.
Sigma-Aldrich
6-Mercaptopurine monohydrate, 98%
Cytarabine, European Pharmacopoeia (EP) Reference Standard
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Dexamethasone for peak identification, European Pharmacopoeia (EP) Reference Standard
Methotrexate for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Vincristine sulfate, meets USP testing specifications
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Vincristine sulfate, European Pharmacopoeia (EP) Reference Standard
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Supelco
Dexamethasone, VETRANAL®, analytical standard
Sigma-Aldrich
Dexamethasone, tested according to Ph. Eur.
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
Methotrexate hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
Methotrexate hydrate, ≥99.0% (sum of enantiomers, HPLC)
Dexamethasone, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate solution, 1.0 mg/mL in methanol with 0.1N NaOH, ampule of 1 mL, certified reference material, Cerilliant®
Methotrexate, European Pharmacopoeia (EP) Reference Standard
SAFC
Methotrexate
Dexamethasone for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Methotrexate, Pharmaceutical Secondary Standard; Certified Reference Material
Methotrexate for peak identification, European Pharmacopoeia (EP) Reference Standard
Supelco
Dexamethasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Dexamethasone-Water Soluble, suitable for cell culture, BioReagent
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%