Merck
  • Ecotoxicological studies of micro- and nanosized barium titanate on aquatic photosynthetic microorganisms.
Ecotoxicological studies of micro- and nanosized barium titanate on aquatic photosynthetic microorganisms.
Aquatic toxicology (Amsterdam, Netherlands) (2014-05-28)
Hudson C Polonini, Humberto M Brandão, Nádia R B Raposo, Ludovic Mouton, Claude Yéprémian, Alain Couté, Roberta Brayner
ABSTRACT
The interaction between live organisms and micro- or nanosized materials has become a current focus in toxicology. As nanosized barium titanate has gained momentum lately in the medical field, the aims of the present work are: (i) to assess BT toxicity and its mechanisms on the aquatic environment, using two photosynthetic organisms (Anabaena flos-aquae, a colonial cyanobacteria, and Euglena gracilis, a flagellated euglenoid); (ii) to study and correlate the physicochemical properties of BT with its toxic profile; (iii) to compare the BT behavior (and Ba(2+) released ions) and the toxic profile in synthetic (Bold's Basal, BB, or Mineral Medium, MM) and natural culture media (Seine River Water, SRW); and (iv) to address whether size (micro, BT MP, or nano, BT NP) is an issue in BT particles toxicity. Responses such as growth inhibition, cell viability, superoxide dismutase (SOD) activity, adenosine-5-triphosphate (ATP) content and photosynthetic efficiency were evaluated. The main conclusions are: (i) BT have statistically significant toxic effects on E. gracilis growth and viability even in small concentrations (1μgmL(-1)), for both media and since the first 24 h; on the contrary of on A. flos-aquae, to whom the effects were noticeable only for the higher concentrations (after 96 h: ≥75 μg mL(-1) for BT NP and =100 μg mL(-1) for BT MP, in BB; and ≥75 μg mL(-1) for both materials in SRW), in spite of the viability being affected in all concentrations; (ii) the BT behaviors in synthetic and natural culture media were slightly different, being the toxic effects more pronounced when grown in SRW - in this case, a worse physiological state of the organisms in SRW can occur and account for the lower resistance, probably linked to a paucity of nutrients or even a synergistic effect with a contaminant from the river; and (iii) the effects seem to be mediated by induced stress without a direct contact in A. flos-aquae and by direct endocytosis in E. gracilis, but in both organisms the contact with both BT MP and BT NP increased SOD activity and decreased photosynthetic efficiency and intracellular ATP content; and (iv) size does not seem to be an issue in BT particles toxicity since micro- and nano-particles produced significant toxic for the model-organisms.
Sigma-Aldrich
Titanium, evaporation slug, diam. × L 6.3 mm × 6.3 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, powder, −100 mesh, 99.7% trace metals basis
Sigma-Aldrich
Titanium, sponge, 1-20 mm, 99.5% trace metals basis
Titanium, IRMM®, certified reference material, 0.1 mm foil
Sigma-Aldrich
Titanium, foil, thickness 0.25 mm, 99.99% trace metals basis
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.127 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Titanium, microfoil, disks, 25mm, thinness 1.0μm, specific density 429μg/cm2, permanent mylar 3.5μm support, 99.6+%
Titanium, tube, 1000mm, outside diameter 0.81mm, inside diameter 0.61mm, wall thickness 0.1mm, as drawn, 99.6+%
Titanium, tube, 100mm, outside diameter 9.5mm, inside diameter 8.2mm, wall thickness 0.65mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 6.1mm, inside diameter 5.1mm, wall thickness 0.5mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 3.0mm, annealed, 99.6+%
Titanium, rod, 1000mm, diameter 2mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 2.03mm, inside diameter 1.63mm, wall thickness 0.2mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 1.5mm, annealed, 99.6+%
Titanium, rod, 50mm, diameter 16mm, as drawn, 99.99+%
Titanium, rod, 1000mm, diameter 6mm, annealed, 99.6+%
Titanium, rod, 100mm, diameter 20mm, as drawn, 99.99+%
Titanium, microfoil, disks, 25mm, thinness 0.1μm, specific density 42.8μg/cm2, permanent mylar 3.5μm support, 99.6+%
Titanium, rod, 1000mm, diameter 5mm, annealed, 99.6+%
Titanium, tube, 200mm, outside diameter 3.0mm, inside diameter 1.5mm, wall thickness 0.75mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 10.3mm, inside diameter 8.7mm, wall thickness 0.8mm, annealed, 99.6+%
Titanium, rod, 500mm, diameter 3.0mm, annealed, 99.6+%
Titanium, tube, 100mm, outside diameter 6.35mm, inside diameter 4.57mm, wall thickness 0.89mm, annealed, 99.6+%
Titanium, tube, 1000mm, outside diameter 3.0mm, inside diameter 1.5mm, wall thickness 0.75mm, annealed, 99.6+%
Titanium, rod, 200mm, diameter 6mm, annealed, 99.6+%
Titanium, sphere, 100pcs, diameter 3mm, 99.6+%
Titanium, tube, 200mm, outside diameter 12.7mm, inside diameter 10.9mm, wall thickness 0.9mm, annealed, 99.6+%