Merck
  • Influence of particle size distribution, organic carbon, pH and chlorides on washing of mercury contaminated soil.
Influence of particle size distribution, organic carbon, pH and chlorides on washing of mercury contaminated soil.
Chemosphere (2014-05-31)
Jingying Xu, Dan B Kleja, Harald Biester, Anders Lagerkvist, Jurate Kumpiene
ABSTRACT
Feasibility of soil washing to remediate Hg contaminated soil was studied. Dry sieving was performed to evaluate Hg distribution in soil particle size fractions. The influence of dissolved organic matter and chlorides on Hg dissolution was assessed by batch leaching tests. Mercury mobilization in the pH range of 3-11 was studied by pH-static titration. Results showed infeasibility of physical separation via dry sieving, as the least contaminated fraction exceeded the Swedish generic guideline value for Hg in soils. Soluble Hg did not correlate with dissolved organic carbon in the water leachate. The highest Hg dissolution was achieved at pH 5 and 11, reaching up to 0.3% of the total Hg. The pH adjustment was therefore not sufficient for the Hg removal to acceptable levels. Chlorides did not facilitate Hg mobilization under acidic pH either. Mercury was firmly bound in the studied soil thus soil washing might be insufficient method to treat the studied soil.
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, for GC derivatization, LiChropur
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
SKU
Pack Size
Availability
Price
Quantity
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mercury, ACS reagent, 99.9995% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether