Merck
  • Anethole and eugenol reduce in vitro and in vivo leukocyte migration induced by fMLP, LTB4, and carrageenan.
Anethole and eugenol reduce in vitro and in vivo leukocyte migration induced by fMLP, LTB4, and carrageenan.
Journal of natural medicines (2014-05-03)
Camila Fernanda Estevão-Silva, Raquel Kummer, Fernanda Carolina Fachini-Queiroz, Renata Grespan, Gessilda Alcântara Nogueira de Melo, Silmara Baroni, Roberto Kenji Nakamura Cuman, Ciomar Aparecida Bersani-Amado
ABSTRACT
The aim of this study was to investigate the effect of anethole (AN) and eugenol (EUG) on leukocyte migration using in vitro chemotaxis and in situ microcirculation assays. BALB/c mice were used for the in vitro chemotaxis assay, and Wistar rats for the in situ microcirculation assay. We evaluated (a) the in vitro leukocyte migration in response to chemotactic factors (formyl-methionyl-leucyl-phenylalanine [fMLP] and leukotriene B4 [LTB4]) and (b) the rolling, adhesion, and migration of leukocytes induced by an injection of carrageenan (100 µg/cavity) into the scrotum of the animal. In the in vitro chemotaxis assay, AN and EUG at doses of 1, 3, 9, and 27 µg/ml significantly inhibited leukocyte migration when stimulated by the chemotactic agents fMLP and LTB4. In the in situ microcirculation assay, AN at doses of 125 and 250 mg/kg and EUG at a dose of 250 mg/kg significantly decreased the number of leukocytes that rolled, adhered, and migrated to perivascular tissue. The results indicate that AN and EUG exert inhibitory effects on leukocyte migration, highlighting their possible use to diminish excessive leukocyte migration in the inflammatory process.
Product Description
Supelco
trans-Anethole, analytical standard
Sigma-Aldrich
Eugenol, ReagentPlus®, 99%
Sigma-Aldrich
Eugenol, natural, ≥98%, FG
Supelco
Eugenol, PESTANAL®, analytical standard
Sigma-Aldrich
trans-Anethole, ≥99%, FCC, FG
Sigma-Aldrich
Eugenol, ≥98%, FCC, FG
Supelco
trans-Anethole, Pharmaceutical Secondary Standard; Certified Reference Material
Eugenol, European Pharmacopoeia (EP) Reference Standard
Supelco
Eugenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
trans-Anethole, 99%
Sigma-Aldrich
Leukotriene B4, ≥97%
Supelco
Eugenol, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, BioXtra, ≥99.0% (TLC)
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, ≥97% (HPLC)
Sigma-Aldrich
Anethol, natural, 99%, FG