Merck
  • A tamoxifen derivative, N,N-diethyl-2-[4-(phenylmethyl) phenoxy] ethanamine, selectively targets P-glycoprotein-positive multidrug resistant Chinese hamster cells.
A tamoxifen derivative, N,N-diethyl-2-[4-(phenylmethyl) phenoxy] ethanamine, selectively targets P-glycoprotein-positive multidrug resistant Chinese hamster cells.
Biochemical pharmacology (2014-05-14)
Elias Georges, Jing Lian, Remi Laberge
ABSTRACT
DPPE, a tamoxifen derivative with antihistamine activity, was previously shown to potentiate the toxicity of chemotherapeutic drugs. Recently, a Phase III clinical study using doxorubicin with DPPE demonstrated significant increase in the overall survival of breast cancer patients. In this study we examined the effects of DPPE alone on the growth of drug sensitive and P-gp positive CHO cell line. Our results demonstrate DPPE is selectively toxic to P-gp positive cells and the sensitivity to DPPE alone correlated with the levels of P-gp expression. Moreover, in MDR cells, DPPE-induced apoptosis was significantly reduced with Bcl2 overexpression and in the presence of P-gp ATPase inhibitor, PSC833. Furthermore, knockdown of P-gp expression in MDR cells with P-gp-siRNA reversed DPPE sensitivity and increased their sensitivity to doxorubicin and taxol but not to cisplatin. The addition of DPPE to membrane fractions led to dose-dependent increase in P-gp ATPase that was inhibited with PSC833. Moreover, incubation of P-gp positive cells with DPPE led to a significant increase in superoxide levels and a drop in cellular ATP and GSH pools that were reversible with inhibitors of P-gp ATPase. The combined presence of DPPE and the mitochondria electron transport complex III inhibitor, antimycin A, synergized in their effects on the growth of MDR cells but had no effect on the growth of parental drug sensitive cells. Collectively, the results of this study provide a possible mechanism that may be relevant to the clinical results of DPPE in breast cancer trial and demonstrates DPPE as P-gp collateral sensitivity drug.
Product Description
Doxorubicin hydrochloride, European Pharmacopoeia (EP) Reference Standard
Paclitaxel semi-synthetic for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Monochlorobimane, suitable for fluorescence, ≥70.0% (HPCE)
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Glutathione, European Pharmacopoeia (EP) Reference Standard
USP
Doxorubicin hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Valspodar, ≥98% (HPLC)
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
Paclitaxel, from Taxus yannanensis, powder
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Paclitaxel, from semisynthetic, ≥98%
Sigma-Aldrich
Paclitaxel, from Taxus brevifolia, ≥95% (HPLC), powder
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
Chlorobimane
Paclitaxel natural for peak identification, European Pharmacopoeia (EP) Reference Standard
Paclitaxel semi-synthetic for system suitability, European Pharmacopoeia (EP) Reference Standard
Paclitaxel, European Pharmacopoeia (EP) Reference Standard