Merck
  • Neurokinin B signaling in the female rat: a novel link between stress and reproduction.
Neurokinin B signaling in the female rat: a novel link between stress and reproduction.
Endocrinology (2014-04-09)
P Grachev, X F Li, M H Hu, S Y Li, R P Millar, S L Lightman, K T O'Byrne
ABSTRACT
Acute systemic stress disrupts reproductive function by inhibiting pulsatile gonadotropin secretion. The underlying mechanism involves stress-induced suppression of the GnRH pulse generator, the functional unit of which is considered to be the hypothalamic arcuate nucleus kisspeptin/neurokinin B/dynorphin A neurons. Agonists of the neurokinin B (NKB) receptor (NK3R) have been shown to suppress the GnRH pulse generator, in a dynorphin A (Dyn)-dependent fashion, under hypoestrogenic conditions, and Dyn has been well documented to mediate several stress-related central regulatory functions. We hypothesized that the NKB/Dyn signaling cascade is required for stress-induced suppression of the GnRH pulse generator. To investigate this ovariectomized rats, iv administered with Escherichia coli lipopolysaccharide (LPS) following intracerebroventricular pretreatment with NK3R or κ-opioid receptor (Dyn receptor) antagonists, were subjected to frequent blood sampling for hormone analysis. Antagonism of NK3R, but not κ-opioid receptor, blocked the suppressive effect of LPS challenge on LH pulse frequency. Neither antagonist affected LPS-induced corticosterone secretion. Hypothalamic arcuate nucleus NKB neurons project to the paraventricular nucleus, the major hypothalamic source of the stress-related neuropeptides CRH and arginine vasopressin (AVP), which have been implicated in the stress-induced suppression of the hypothalamic-pituitary-gonadal axis. A separate group of ovariectomized rats was, therefore, used to address the potential involvement of central CRH and/or AVP signaling in the suppression of LH pulsatility induced by intracerebroventricular administration of a selective NK3R agonist, senktide. Neither AVP nor CRH receptor antagonists affected the senktide-induced suppression of the LH pulse; however, antagonism of type 2 CRH receptors attenuated the accompanying elevation of corticosterone levels. These data indicate that the suppression of the GnRH pulse generator by acute systemic stress requires hypothalamic NKB/NK3R signaling and that any involvement of CRH therewith is functionally upstream of NKB.
Sigma-Aldrich
Corticosterone, ≥98.5% (HPLC)
Supelco
Corticosterone, VETRANAL®, analytical standard
Sigma-Aldrich
Corticosterone, ≥92%
Sigma-Aldrich
Corticotropin Releasing Factor sheep, ≥95% (HPLC)
Sigma-Aldrich
Neurokinin B, ≥95% (HPLC)
Sigma-Aldrich
Xylazine, ≥99%
Sigma-Aldrich
Astressin, ≥90% (HPLC)
Gonadorelin, European Pharmacopoeia (EP) Reference Standard
Carprofen for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, for molecular biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, anhydrous, ≥99.9%
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Supelco
Dimethyl sulfoxide, analytical standard
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, for molecular biology
Supelco
Carprofen, VETRANAL®, analytical standard
Supelco
Corticosterone solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Dimethyl sulfoxide, European Pharmacopoeia (EP) Reference Standard
Supelco
Carprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Carprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Dimethyl sulfoxide, for inorganic trace analysis, ≥99.99995% (metals basis)
Sigma-Aldrich
Senktide trifluoroacetate salt, ≥95% (HPLC)
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, suitable for HPLC, ≥99.7%
USP
Dimethyl sulfoxide, United States Pharmacopeia (USP) Reference Standard