Merck
Arabidopsis ein2-1 and npr1-1 response to Al stress.
Bulletin of environmental contamination and toxicology (2014-03-13)
Yiyan Zhang, Qingqing He, Shiyang Zhao, Linli Huang, Lin Hao
ABSTRACT
An enhanced Al(3+) tolerance has been observed in ethylene insensitive mutant ein2-1 and salicylic acid insensitive mutant npr1-1 of Arabidopsis. However, we found that the tolerant phenotype of ein2-1 and npr1-1 under Al stress was dependent on NPR and EIN function, respectively, because the double mutant ein2-1/npr1-1 displayed more sensitive to Al stress than wild-type plants. We analysed the differential performance between ein2-1/npr1-1 and their respective single mutant in response to Al stress, and found that antioxidant defence rather than malate exudation was the determinant factor.
Product Description
Aluminum, IRMM®, certified reference material, 1.0 mm foil
Sigma-Aldrich
Aluminum, evaporation slug, diam. × L 6.3 mm × 6.3 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 0.58 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Aluminum, pellets, 3-8 mesh, ≥99.999% trace metals basis
Sigma-Aldrich
Aluminum, wire, diam. 1.0 mm, 99.999% trace metals basis
Sigma-Aldrich
Aluminum, granular, <1 mm, 99.7% trace metals basis
Aluminum, IRMM®, certified reference material, 1.0 mm wire
Sigma-Aldrich
Aluminum, foil, thickness 8 μm, 99% trace metals basis
Aluminum, rod, 50mm, diameter 28mm, as drawn, 99.999%
Aluminum, rod, 100mm, diameter 6.0mm, as drawn, 99.999%
Aluminum, microfoil, disks, 50mm, thinness 1.0μm, specific density 269.9μg/cm2, permanent mylar 3.5μm support, 99.999%
Aluminum, tube, 100mm, outside diameter 0.55mm, inside diameter 0.4mm, wall thickness 0.075mm, as drawn, 99.5%
Aluminum, rod, 44mm, diameter 77.5mm, as drawn, 99.999%
Aluminum, tube, 200mm, outside diameter 0.86mm, inside diameter 0.76mm, wall thickness 0.05mm, as drawn, 99.5%
Aluminum, microfoil, disks, 25mm, thinness 0.0075μm, specific density 1.84μg/cm2, permanent mylar 3.5μm support, 99.999%
Aluminum, tube, 0.5m coil, as drawn, outside diameter 0.30mm, inside diameter 0.2mm, wall thickness 0.05mm, 99.5%
Aluminum, wire reel, 100m, diameter 0.5mm, as drawn, 99.95+%
Aluminum, wire reel, 100m, diameter 0.063mm, annealed, 99.99%
Aluminum, tube, 100mm, outside diameter 0.6mm, inside diameter 0.3mm, wall thickness 0.15mm, as drawn, 99.5%
Aluminum, mesh, 150x150mm, nominal aperture 0.38mm, wire diameter 0.25mm, 40x40 wires/inch, open area 37%, plain weave mesh
Aluminum, tube, 200mm, outside diameter 1.42mm, inside diameter 1.166mm, wall thickness 0.127mm, as drawn, 99.5%
Aluminum, rod, 200mm, diameter 10.0mm, as drawn, 99.999%
Aluminum, tube, 500mm, outside diameter 0.55mm, inside diameter 0.4mm, wall thickness 0.075mm, as drawn, 99.5%
Aluminum, tube, 500mm, outside diameter 3.0mm, inside diameter 1.62mm, wall thickness 0.69mm, hard, 99.5%
Aluminum, wire reel, 100m, diameter 0.25mm, hard, 99.99+%
Aluminum, sphere, 100pcs, diameter 1.0mm, aluminum alloy 2017 precision sphere, grade 200, 95.2%
Aluminum, wire reel, 100m, diameter 0.025mm, as drawn, 99.0+%
Aluminum, rod, 200mm, diameter 12.7mm, 99%
Aluminum, wire reel, 0.5m, diameter 1.0mm, as drawn, 99.999%
Aluminum, rod, 500mm, diameter 15.0mm, as drawn, 99.999%