Merck
  • A phase III randomized, placebo-controlled study of topical amitriptyline and ketamine for chemotherapy-induced peripheral neuropathy (CIPN): a University of Rochester CCOP study of 462 cancer survivors.
A phase III randomized, placebo-controlled study of topical amitriptyline and ketamine for chemotherapy-induced peripheral neuropathy (CIPN): a University of Rochester CCOP study of 462 cancer survivors.
Supportive care in cancer : official journal of the Multinational Association of Supportive Care in Cancer (2014-02-18)
Jennifer S Gewandter, Supriya G Mohile, Charles E Heckler, Julie L Ryan, Jeffrey J Kirshner, Patrick J Flynn, Judith O Hopkins, Gary R Morrow
ABSTRACT
Chemotherapy-induced peripheral neuropathy (CIPN) occurs in as high as 70% of patients receiving certain types of chemotherapy agents. The FDA has yet to approve a therapy for CIPN. The aim of this multicenter, phase III, randomized, double-blind, placebo-controlled trial was to investigate the efficacy of 2% ketamine plus 4% amitriptyline (KA) cream for reducing CIPN. Cancer survivors who completed chemotherapy at least 1 month prior and had CIPN (>4 out of 10) were enrolled (N=462). CIPN was assessed using average scores from a 7-day daily diary that asks patients to rate the average "pain, numbness, or tingling in [their] hands and feet over the past 24 h" on an 11-point numeric rating scale at baseline and 6 weeks post intervention. ANCOVA was used to measure differences in 6-week CIPN with effects including baseline CIPN, KA treatment arm, and previous taxane therapy (Y/N). The KA treatment showed no effect on 6-week CIPN scores (adjusted mean difference=-0.17, p=0.363). This study suggests that KA cream does not decrease CIPN symptoms in cancer survivors.
Product Description
Supelco
Ketamine hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
USP
Amitriptyline hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Amitriptyline hydrochloride, ≥98% (TLC), powder
Supelco
Amitriptyline hydrochloride solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Ketamine hydrochloride, European Pharmacopoeia (EP) Reference Standard
Amitriptyline hydrochloride, European Pharmacopoeia (EP) Reference Standard
Supelco
Amitriptyline hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material