Merck
  • Anticoagulant medication at time of needle biopsy for breast cancer in relation to risk of lymph node metastasis.
Anticoagulant medication at time of needle biopsy for breast cancer in relation to risk of lymph node metastasis.
International journal of cancer (2013-12-19)
Rickard Ljung, Roland Sennerstam, Fredrik Mattsson, Gert Auer, Jesper Lagergren
ABSTRACT
Anticoagulant treatment might enhance the natural defense against tumor cell dissemination caused by diagnostic needle biopsy by counteracting thrombocyte coating of such cells. To clarify whether women using anticoagulant treatment at the time of biopsy have a lower occurrence of lymph node metastasis, we conducted a nationwide Swedish cohort study of 26,528 female incident breast cancer patients in 2006-2011. Point risk ratio (RR) of risk of lymph node metastasis among users of anticoagulant treatment adjusted for age, T-stage, socioeconomic factors, and concomitant medication was RR = 0.94, (95% CI: 0.87-1.03), and lower in younger women (RR = 0.80, 95% CI 0.50-1.29). Although nonsignificant, these associations may underestimate a true negative association since women using anticoagulant treatment are likely to have more concomitant diseases, lead an unhealthier lifestyle, and have lower participation in mammography screening. These findings provide some support for the hypothesis that anticoagulant medications might counteract breast cancer spread caused by needle biopsy.
Product Description
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Aspirin, meets USP testing specifications
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%