Merck
  • Hexanuclear copper(II) cages built on a central {μ3-O···H···μ3-O} moiety, 1,3-bis(dimethylamino)-2-propanolato and capping R-phosphonates: crystal structures, magnetic behavior, and DFT studies.
Hexanuclear copper(II) cages built on a central {μ3-O···H···μ3-O} moiety, 1,3-bis(dimethylamino)-2-propanolato and capping R-phosphonates: crystal structures, magnetic behavior, and DFT studies.
Inorganic chemistry (2012-05-26)
Saskia Speed, Ramon Vicente, Daniel Aravena, Eliseo Ruiz, Olivier Roubeau, Simon J Teat, M Salah El Fallah
ABSTRACT
The syntheses, structural characterization, and magnetic behavior of two new hexanuclear copper(II) complexes derived from R-phosphonic acids and 1,3-bis(dimethylamino)-2-propanol (Hbdmap) with formulas [Cu(6)(μ-bdmap)(3)(μ(3)-Ph-PO(3))(2)(μ(3)-O···H···μ(3)-O)(ClO(4))(2)(H(2)O)]·5H(2)O (1) and [Cu(6)(μ-bdmap)(3)(μ(3)-t-Bu-PO(3))(2)(μ(3)-O···H···μ(3)-O)(μ(1,3)-dca)(dca)(H(2)O)]·6H(2)O (2) (Ph-H(2)PO(3) = phenylphosphonic acid, t-Bu-H(2)PO(3) = tert-butylphosphonic acid, dca = dicyanamide) are reported. Compounds 1 and 2 are hexanuclear 3.111 R-phosphonate(2-)/1,3-bis(dimethylamino)-2-propanolato(1-) cages including in the center the [μ(3)-O···H···μ(3)-O](3-) unit. The temperature dependence of the magnetic properties of 1 and 2 clearly indicates an overall strong antiferromagnetic coupling confirmed by DFT calculations.
Sigma-Aldrich
tert-Butylphosphonic acid, 98%