Merck
Pharmacodynamics and pharmacokinetics of thiopental.
Clinical pharmacokinetics (1998-09-18)
H Russo, F Bressolle
ABSTRACT
Thiopental is an ultra short-acting barbiturate which remains the standard against which other induction agents are judged; it is also indicated for the therapy of brain hypoxic-ischaemia injuries and status epilepticus. Aspects of drug distribution that govern the onset and end of drug effect have been intensively studied to determine which parameters (in patient characteristics, diseases and administration modalities) influence effective dose and concentrations in individual patients. Thiopental has been used as a reference for pharmacokinetic and/or pharmacodynamic models in the study of rapid and short acting effect drugs. In anaesthesiology the pharmacokinetics of thiopental are described as linear; when doses and duration of treatment increase, nonlinear pharmacokinetics occur because of the saturation and/or the induction of the metabolism.
Product Description
Supelco
(±)Thiopental solution, analytical standard, for drug analysis