Merck
  • Fosfomycin tromethamine. A review of its antibacterial activity, pharmacokinetic properties and therapeutic efficacy as a single-dose oral treatment for acute uncomplicated lower urinary tract infections.
Fosfomycin tromethamine. A review of its antibacterial activity, pharmacokinetic properties and therapeutic efficacy as a single-dose oral treatment for acute uncomplicated lower urinary tract infections.
Drugs (1997-04-01)
S S Patel, J A Balfour, H M Bryson
ABSTRACT
Fosfomycin tromethamine is a phosphonic acid bactericidal agent with in vitro activity against most urinary tract pathogens. It is particularly active against Escherichia coli, and Citrobacter, Enterobacter, Klebsiella, Serratia and Enterococcus spp. There appears to be little cross-resistance between fosfomycin and other antibacterial agents, possibly because it differs from other agents in its general chemical structure and site of action. In its new formulation as the oral tromethamine salt, fosfomycin has 34 to 41% oral bioavailability, has a mean elimination half-life of 5.7 hours, and is primarily excreted unchanged in the urine. Following a single 3 g oral dose, peak urinary concentrations occur within 4 hours and remain high (> 128 mg/L) for 24 to 48 hours, which is sufficient to inhibit most urinary tract pathogens. In clinical trials in patients with acute uncomplicated lower urinary tract infection, single-dose fosfomycin tromethamine therapy was effective, and comparable with several other antibacterial agents given either as single-dose or multiple-dose treatments [e.g. beta-lactam and fluoroquinolone agents, cotrimoxazole (trimethoprim-sulfamethoxazole), nitrofurantoin and pipemidic acid]. Bacteriological eradication rates of 75 to 90% were achieved 5 to 11 days after therapy, with eradication rates of 62 to 93% 4 to 6 weeks after therapy. In 3 large double-blind comparisons with ciprofloxacin, cotrimoxazole and nitrofurantoin, 99% of fosfomycin tromethamine recipients and 100% of patients receiving comparator agents were considered clinically cured or improved after therapy. Fosfomycin tromethamine is well tolerated, with a low incidence of adverse events. These comprise mainly gastrointestinal symptoms that are transient, mild and self-limiting. Thus, fosfomycin tromethamine achieves high clinical and bacteriological cure rates in patients with acute uncomplicated lower urinary tract infection and is well tolerated. The single-dose administration regimen and favourable US pregnancy category rating of fosfomycin tromethamine should also encourage its use in this indication.
Sigma-Aldrich
Tris(hydroxymethyl)aminomethane, ACS reagent, ≥99.8%
Sigma-Aldrich
Trizma® base, puriss. p.a., ≥99.7% (T)
Sigma-Aldrich
Trizma® base, ≥99.0% (T)
Sigma-Aldrich
Trizma® base, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Trizma® base, BioUltra, for molecular biology, ≥99.8% (T)
Sigma-Aldrich
Trizma® base, anhydrous, free-flowing, Redi-Dri, ≥99.9%
Sigma-Aldrich
Trizma® base, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Trizma® base, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)
Sigma-Aldrich
Trizma® base, ≥99.9% (titration), crystalline
Supelco
Trizma® base, reference material for titrimetry, certified by BAM, >99.5%
Trometamol, European Pharmacopoeia (EP) Reference Standard
Supelco
Tromethamine, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), crystalline
SAFC
Tromethamine