Merck
Norethindrone--a review of therapeutic applications.
International journal of fertility and menopausal studies (1995-07-01)
H F Taitel, M E Kafrissen
ABSTRACT
Norethindrone is utilized for numerous noncontraceptive therapies, especially during the menopause. Through an analysis of the literature we have described the pharmacological profile of norethindrone and the potential therapeutic implications of low-dose therapies, with an emphasis on endometrial pathology, climacteric symptoms, lipid metabolism, and bone density.
Product Description
Norethisterone for system suitability, European Pharmacopoeia (EP) Reference Standard
USP
Norethindrone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
19-Norethindrone, ≥98%, powder
Norethisterone, European Pharmacopoeia (EP) Reference Standard