Merck
  • Comparative efficacy and safety of cefprozil versus penicillin, cefaclor and erythromycin in the treatment of streptococcal pharyngitis and tonsillitis.
Comparative efficacy and safety of cefprozil versus penicillin, cefaclor and erythromycin in the treatment of streptococcal pharyngitis and tonsillitis.
European journal of clinical microbiology & infectious diseases : official publication of the European Society of Clinical Microbiology (1994-10-01)
J M McCarty
ABSTRACT
Cefprozil is a new oral cephalosporin with an enhanced in vitro spectrum of activity that includes group A beta-hemolytic streptococci (GABHS). Four multicenter randomized clinical trials were conducted to compare the clinical efficacy and safety of cefprozil administered once or twice a day for the treatment of mild to moderate GABHS tonsillitis and pharyngitis. A total of 1597 patients were enrolled in the trials. Patient demographics and severity of infection were similar for all treatment groups. In Study 1, cefprozil administered at 20 mg/kg once daily was clinically, in 68 of 76 patients (89%) and bacteriologically, in 66 of 74 patients (89%) superior to penicillin -51 of 69 (74%) and 46 of 69 (67%)--administered three times a day in patients of two to 12 years of age. In Study 2, the patients enrolled were 13 years of age and older, and cefprozil administered at 20 mg/kg once a day had similar clinical (93% vs. 90%) and bacteriological (95% vs. 94%) response rates as cefaclor administered three times a day. Study 3 demonstrated that cefprozil administered twice daily was similar to penicillin given three times a day, the clinical satisfactory response being 164 of 175 (94%) for cefprozil and 146 of 165 (88%) for penicillin. In Study 4, identical clinical and bacteriologic responses (95%) were observed for cefprozil administered once a day and erythromycin ethylsuccinate administered four times a day in children over two years of age. There were no significant differences in the incidence or severity of drug-related adverse events, which, when reported, were mild and transient.(ABSTRACT TRUNCATED AT 250 WORDS)
Sigma-Aldrich
Cefaclor