Merck
  • Epsilon aminocaproic acid in hemophiliacs undergoing dental extractions: a concise review.
Epsilon aminocaproic acid in hemophiliacs undergoing dental extractions: a concise review.
Oral surgery, oral medicine, and oral pathology (1981-02-01)
O N Lucas, T W Albert
ABSTRACT
On the basis of our review of the literature, it can be concluded that the advantages of E-A.C.A. include (1) safe execution of oral surgical procedures in persons with hemophilia of varying degrees of severity, (2) decrease or total elimination of the need for specific factor replacement therapy, (3) extremely positive cost effectiveness, and (4) reproducibility of results at different hemophilia centers.
Sigma-Aldrich
6-Aminocaproic acid, ≥99% (titration), powder
Sigma-Aldrich
6-Aminohexanoic acid, ≥98.5% (NT)
Aminocaproic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Aminocaproic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
6-Aminocaproic acid, BioUltra, ≥99%