Merck
Disposition and response to bumetanide and furosemide.
The American journal of cardiology (1986-01-24)
D C Brater
ABSTRACT
Bumetanide and furosemide are potent loop diuretics; the former is 40 to 50 times more potent than the latter on a weight basis. Bumetanide is absorbed more quickly than furosemide and is twice as bioavailable. Both drugs exhibit changes in elimination in the presence of renal insufficiency as well as changes in the time course of absorption in congestive heart failure. More data are needed to assess potential differences between them in various clinical conditions.
Product Description
Bumetanide, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Bumetanide, ≥98%