Sigma-Aldrich
Nickel, wire, diam. 0.25 mm, ≥99.9%
Sigma-Aldrich
Nickel, foil, thickness 0.125 mm, ≥99.9%
Sigma-Aldrich
Nickel, foil, thickness 0.25 mm, 99.995% trace metals basis
Sigma-Aldrich
Nickel, wire, diam. 0.5 mm, ≥99.99% trace metals basis
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Sigma-Aldrich
Nickel, powder, <150 μm, 99.99% trace metals basis
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 3202, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 4200, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2800, slurry, in H2O, active catalyst
Sigma-Aldrich
Raney®-Nickel, W.R. Grace and Co. Raney® 2400, slurry, in H2O, active catalyst
Nickel, tube, 200mm, outside diameter 1.3mm, inside diameter 1.1mm, wall thickness 0.1mm, hard, 99.5%
Nickel, tube, 500mm, outside diameter 0.8mm, inside diameter 0.3mm, wall thickness 0.25mm, as drawn, 99.5%
Nickel, foil, 25mm disks, thickness 0.008mm, 99.95%
Nickel, tube, 500mm, outside diameter 0.762mm, inside diameter 0.204mm, wall thickness 0.279mm, hard, 99.5%
Nickel, tube, 200mm, outside diameter 2.5mm, inside diameter 2.25mm, wall thickness 0.125mm, hard, 99.5%
Nickel, wire reel, 5m, diameter 0.4mm, as drawn, 99.99+%
Nickel, wire reel, 50m, diameter 0.4mm, as drawn, 99%
Nickel, wire reel, 2m, diameter 0.5mm, hard, 99.98%
Nickel, foil, 25mm disks, thickness 0.0025mm, 99.95%
Nickel, wire reel, 5m, diameter 0.75mm, as drawn, 99%
Nickel, wire reel, 50m, diameter 2.0mm, hard, 99.98%
Nickel, wire reel, 200m, diameter 0.125mm, annealed, 99.98%
Nickel, tube, 200mm, outside diameter 6.5mm, inside diameter 5.5mm, wall thickness 0.5mm, hard, 99.5%
Nickel, wire reel, 20m, diameter 0.125mm, as drawn, 99%
Nickel, foil, 25mm disks, thickness 0.009mm, 99.95%
Nickel, tube, 200mm, outside diameter 2.5mm, inside diameter 2mm, wall thickness 0.25mm, hard, 99.5%
Nickel, tube, 500mm, outside diameter 0.78mm, inside diameter 0.2mm, wall thickness 0.29mm, as drawn, 99.5%
Nickel, tube, 200mm, outside diameter 0.8mm, inside diameter 0.3mm, wall thickness 0.25mm, as drawn, 99.5%
Nickel, wire reel, 250m, diameter 0.125mm, as drawn, 99%
Nickel, wire reel, 5m, diameter 0.25mm, as drawn, 99%