Merck
Xylitol.
Topics in companion animal medicine (2013-06-26)
Michael E Peterson
ABSTRACT
Xylitol is a prevalent sugar substitute found in a wide variety of foods, particularly those labeled as "low carb." It is found in many medicines and dental products both for its antibacterial activity and to increase palatability. Originally, this toxin was recognized as a problem in dogs following sugarless gum ingestions. Xylitol is generally nontoxic to mammals except for dogs. In the dog, xylitol induces marked increases in insulin production and occasionally hepatopathy. The clinical syndrome is manifested with signs consistent with profound hypoglycemia, hypokalemia, hypophosphatemia, and acute hepatic failure. Treatment relies upon administration of intravenous glucose, hepatic support, and general supportive care.
Sigma-Aldrich
Xylitol, ≥99% (GC)
Sigma-Aldrich
Xylitol
Xylitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Xylitol, Pharmaceutical Secondary Standard; Certified Reference Material